Skip to Content
Merck
All Photos(2)

Key Documents

540978

Sigma-Aldrich

2,2-Dibromo-2-cyanoacetamide

96%

Synonym(s):

Dibromocyano acetic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2C(CN)CONH2
CAS Number:
Molecular Weight:
241.87
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

mp

122-125 °C (lit.)

functional group

amide
bromo
nitrile

SMILES string

NC(=O)C(Br)(Br)C#N

InChI

1S/C3H2Br2N2O/c4-3(5,1-6)2(7)8/h(H2,7,8)

InChI key

UUIVKBHZENILKB-UHFFFAOYSA-N

General description

2,2-Dibromo-2-cyanoacetamide, also known as 2,2-dibromo-3-nitrilopropionamide (DBNPA), can be synthesized by reacting sodium bromide and cyanoacetamide. Its crystals are monoclinic and belong to the space group P21/n.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Fernanda Campa et al.
Applied and environmental microbiology, 85(21) (2019-08-25)
Production of unconventional oil and gas continues to rise, but the effects of high-density hydraulic fracturing (HF) activity near aquatic ecosystems are not fully understood. A commonly used biocide in HF, 2,2-dibromo-3-nitrilopropionamide (DBNPA), was studied in microcosms of HF-impacted (HF+)
Synthesis of 2, 2-dibromo-2-cyanoacetamide by in situ oxidation bromination.
LI WX and LI FG.
Fine and Specialty Chemicals / Jing Xi Yu Zhuan Yong Hua Xue Pin, 11, 018-018 (2012)
A Senoh et al.
Clinical and experimental dermatology, 34(8), e732-e736 (2009-08-12)
Erythema multiforme (EM)-like contact reactions are induced by exposure to various chemicals. We report two industrial workers from a paper mill who presented with generalized EM-like lesions 17 and 10 days, respectively, after accidental exposure to liquid-formulated 2,2-dibromo-3-nitrilopropionamide (DBNPA) 20%.
Anna Rigol et al.
Environmental toxicology and chemistry, 23(2), 339-347 (2004-02-26)
The risk associated with wood extractives, biocides, and other additives in pulp and paper mill effluents was evaluated by performing a characterization of process waters and effluents in terms of toxicity and chemical analysis. The individual toxicity of 10 resin
Fang Chen
Toxicology and industrial health, 28(2), 181-185 (2011-09-23)
The chronic aquatic toxicities of a microbicide dibromonitrilopropionamide (DBNPA) in Daphnia magna and rainbow trout were evaluated. DBNPA can significantly affect the reproduction and survival of D. magna. The lowest observed effective concentration (LOEC) and the no observed effective concentration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service