540757
16-Hexadecanolide
97%
Synonym(s):
Juniperic acid lactone, NSC 33546
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C16H30O2
CAS Number:
Molecular Weight:
254.41
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
refractive index
n20/D 1.4680-1.4730
mp
34-38 °C (lit.)
functional group
ester
SMILES string
O=C1CCCCCCCCCCCCCCCO1
InChI
1S/C16H30O2/c17-16-14-12-10-8-6-4-2-1-3-5-7-9-11-13-15-18-16/h1-15H2
InChI key
LOKPJYNMYCVCRM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
16-Hexadecanolide (HDL) is a 7-membered lactone. It can be synthesized from 16-hydroxyhexadecanoic acid ethyl ester. HDL undergoes enzymatic ring-opening polymerization in the presence of lipases to afford the corresponding polymers.
Application
16-Hexadecanolide may be used as a starting reagent in the synthesis of caeliferin A 16:0.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A Facile and Efficient Synthesis of Caeliferin a 16: 0.
Peng C, et al.
Chemistry of Natural Compounds, 2(52), 199-201 (2016)
Lactone synthesis from 16-hydroxyhexadecanoic acid ethyl ester in organic solvents catalyzed with polyethylene glycol-modified lipase.
Kodera Y, et al.
Journal of Biotechnology, 31(2), 219-224 (1993)
Lipase-catalyzed ring-opening polymerization of 16-hexadecanolide.
Namekawa S, et al.
Proceedings of the Japan Academy. Series B, Physical and Biological Sciences, 74(4), 65-68 (1998)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service