All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H16Si
CAS Number:
Molecular Weight:
116.28
Beilstein:
1731336
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥97.0%
refractive index
n20/D 1.414
density
0.721 g/mL at 20 °C (lit.)
SMILES string
CCCCCC[SiH3]
InChI
1S/C6H16Si/c1-2-3-4-5-6-7/h2-6H2,1,7H3
InChI key
QGGUMTNPIYCTSF-UHFFFAOYSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
53.6 °F - closed cup
Flash Point(C)
12 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tang Dang et al.
Biosensors & bioelectronics, 142, 111494-111494 (2019-07-19)
Almost no analytical assays, either colorimetric or fluorescence assays, for generic microplate readers is capable of dynamic measurements of protein-protein binding or the quantification of kinetic association and dissociation constants of protein interactions. On the other hand, protein binding kinetics
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service