Skip to Content
Merck
All Photos(1)

Key Documents

52340

Sigma-Aldrich

Hexadecylpyridinium bromide

≥97.0%

Synonym(s):

Cetylpyridinium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H38BrN
CAS Number:
Molecular Weight:
384.44
Beilstein:
3778948
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

description

cationic

Assay

≥97.0%

mol wt

384.44 g/mol

technique(s)

titration: suitable

SMILES string

[Br-].CCCCCCCCCCCCCCCC[n+]1ccccc1

InChI

1S/C21H38N.BrH/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-19-22-20-17-15-18-21-22;/h15,17-18,20-21H,2-14,16,19H2,1H3;1H/q+1;/p-1

InChI key

DVBJBNKEBPCGSY-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Hexadecylpyridinium bromide (HDPB) is a cationic surfactant with quaternary nitrogen located in the aromatic heterocyclic ring of the headgroup and a hydrocarbon tail consisting of 16 carbons. It has application in the polymerization reaction as well as in the extraction and preconcentration of ionic species.

Application

Hexadecylpyridinium bromide can be used as a cationic surfactant to:
  • Prepare a polyfluorate salt.
  • Inhibit the corrosion of mild steel in acidic media.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular modeling of cetylpyridinium bromide, a cationic surfactant, in solutions and micelle
Verma R, et al.
Journal of chemical theory and computation, 11(11), 5415-5425 (2015)
Inhibition of mild steel corrosion by hexadecylpyridinium bromide in 0.5 M H2SO4}
Saleh MM
Materials Chemistry and Physics, 98(1), 83-89 (2006)
Cationic surfactant-based polyfluorate salts: Phase separation and analytical applications in the extraction and preconcentration of ionic species prior to liquid chromatography
Paleologos EK
Journal of Chromatography A, 1066(1-2), 19-25 (2005)
Jason Lin et al.
Materials (Basel, Switzerland), 13(8) (2020-04-30)
Biphenyl wrinkled mesoporous silica nanoparticles with controlled particle size and high surface area were evaluated for the storage and delivery of doxorubicin. The average particle size and surface area were ~70 nm and ~1100 m2/g. The doxorubicin loading efficiency was
Soma Sharma et al.
The journal of physical chemistry. B, 119(34), 11295-11306 (2015-07-07)
Reverse micelles as nanoreactors have been most successful in designing nanostructures of different sizes and shapes. Nevertheless, important questions regarding the explicit roles of intrinsic parameters in modifying soft colloid templates which eventually give rise to variety of nanostructures are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service