518379
Butylboronic acid N,N,N′,N′-tetramethyl-L-tartaric acid diamide ester
Synonym(s):
(4R,5R)-2-Butyl-N,N,N′,N′-tetramethyl-1,3,2-dioxaborolane-4,5-dicarboxamide
About This Item
Recommended Products
optical activity
[α]20/D -105°, c = 1.7% in chloroform
Quality Level
refractive index
n20/D 1.478 (lit.)
bp
178-179 °C (lit.)
density
1.072 g/mL at 25 °C (lit.)
functional group
amide
SMILES string
CCCCB1O[C@H]([C@@H](O1)C(=O)N(C)C)C(=O)N(C)C
InChI
1S/C12H23BN2O4/c1-6-7-8-13-18-9(11(16)14(2)3)10(19-13)12(17)15(4)5/h9-10H,6-8H2,1-5H3/t9-,10-/m1/s1
InChI key
AFQWQRBBIZKYTE-NXEZZACHSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- (+)-Frondosin, a natural product from marine sponges.
- Marine cyanobacterium natural products: coibacins A and B.
Preparation of biologically and pharmacologically active molecules
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service