Skip to Content
Merck
All Photos(2)

Key Documents

513474

Sigma-Aldrich

Luperox® A75FP, Benzoyl peroxide

contains 25 wt. % water as stabilizer, 75%

Synonym(s):

Benzoyl peroxide moistened with water, BPO, Benzoyl peroxide, Diacyl peroxide, Dibenzoyl peroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CO)2O2
Molecular Weight:
242.23
Beilstein:
984320
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Product Name

Luperox® A75FP, Benzoyl peroxide, 75% remainder water, contains 25 wt. % water as stabilizer, 75%

Quality Level

Assay

75%

form

powder

contains

25 wt. % water as stabilizer

reaction suitability

reagent type: oxidant

mp

103 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C(OOC(=O)c1ccccc1)c2ccccc2

InChI

1S/C14H10O4/c15-13(11-7-3-1-4-8-11)17-18-14(16)12-9-5-2-6-10-12/h1-10H

InChI key

OMPJBNCRMGITSC-UHFFFAOYSA-N

Other Notes

Self Accelerating Decomposition Temperature (SADT): 71 °C

Legal Information

Luperox is a registered trademark of Arkema Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Org. Perox. C - Skin Sens. 1A

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Boot et al.
European cells & materials, 39, 108-120 (2020-02-20)
Local prophylaxis with antibiotic-loaded bone cement is a successful method to prevent post-operative infections in patients receiving orthopaedic implants. No comparable method is available for uncemented implants. Therefore, a hydrogel consisting of hyaluronic and polylactic acids was evaluated in a
Laszlo Prokai et al.
Science translational medicine, 7(297), 297ra113-297ra113 (2015-07-24)
Many neurological and psychiatric maladies originate from the deprivation of the human brain from estrogens. However, current hormone therapies cannot be used safely to treat these conditions commonly associated with menopause because of detrimental side effects in the periphery. The
Megan E Oest et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 33(3), 334-342 (2014-11-20)
Late-onset fragility fractures are a common complication following radiotherapy for metastatic disease and soft tissue sarcomas. Using a murine hindlimb focal irradiation model (RTx), we quantified time-dependent changes in osteoclasts and mineral apposition rate (MAR). Mice received either a single
Meital Bloch et al.
International journal of pharmaceutics, 478(2), 504-516 (2014-12-02)
To increase colonoscopy capability to discriminate benign from malignant polyps, we suggest combining two imaging approaches based on targeted polymeric platforms. Water-soluble cationized polyacrylamide (CPAA) was tagged with the near infrared (NIR) dye IR-783-S-Ph-COOH to form Flu-CPAA. The recognition peptide
Ahmed M Debela et al.
Biosensors & bioelectronics, 71, 25-29 (2015-04-17)
An alternative strategy for surface tethering of DNA probes, where highly reactive glassy carbon (GC) substrates are prepared via electrochemical hydrogenation and electrochemical/chemical chlorination is reported. Thiolated DNA probes and alkanethiols were stably immobilised on the halogenated carbon, with electrochemical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service