Skip to Content
Merck
All Photos(1)

Key Documents

511188

Sigma-Aldrich

Methyl indole-5-carboxylate

99%

Synonym(s):

Indole-5-carboxylic acid, methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

126-128 °C (lit.)

SMILES string

COC(=O)c1ccc2[nH]ccc2c1

InChI

1S/C10H9NO2/c1-13-10(12)8-2-3-9-7(6-8)4-5-11-9/h2-6,11H,1H3

InChI key

DRYBMFJLYYEOBZ-UHFFFAOYSA-N

General description

Methyl indole-5-carboxylate (Methyl 1H-indole-5-carboxylate), a substituted 1H-indole, can be prepared by the esterification of indole-5-carboxylic acid. Its efficacy as a substrate for indigoid generation has been assessed.

Application

Methyl indole-5-carboxylate may be used as a reactant in the following processes:
  • biosynthesis of inhibitors of protein kinases
  • metal-free Friedel-Crafts alkylation
  • preparation of diphenylsulfonium ylides from Martin′s sulfurane
  • cross dehydrogenative coupling reactions
  • synthesis of indirubin derivatives
  • preparation of aminoindolylacetates
Methyl indole-5-carboxylate may be used in the preparation of:
  • methyl indoline-5-carboxylate
  • 1H-indole-5-carbohydrazide
  • dimethyl 1H-indole-3,5-dicarboxylate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A facial synthesis and antimicrobial activity of some pyrazole derivatives carrying indole.
Sarma KN, et al.
Journal of Chemistry, 7(3), 745-750 (2010)
A direct ylide transfer to carbonyl derivatives and heteroaromatic compounds.
Xueliang Huang et al.
Angewandte Chemie (International ed. in English), 49(47), 8979-8983 (2010-10-13)
Wen-Jie Lu et al.
European journal of medicinal chemistry, 64, 498-511 (2013-05-21)
This report describes the synthesis, and in vitro and in vivo antimalarial evaluations of certain ester-modified neocryptolepine (5-methyl-5H-indolo[2,3-b]quinoline) derivatives. The modifications were carried out by introducing ester groups at the C2 and/or C9 position on the neocryptolepine core and the
Fei Yang et al.
Organic letters, 12(22), 5214-5217 (2010-10-23)
A novel cross dehydrogenative coupling (CDC) reaction of N,N-dimethylanilines with methyl ketones by cooperative copper and aminocatalysis has been developed, which leads to the formation of β-arylamino ketones in 42-73% yields. Moreover, the copper-catalyzed alkylation of free (NH) indoles with
Wang, T. C.; et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 21, 1407-1407 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service