Skip to Content
Merck
All Photos(1)

Documents

499552

Sigma-Aldrich

Zinc preparation

5 g/dL Zn+2 in THF, highly reactive Rieke®metal

Synonym(s):

Zn

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: zinc
reagent type: catalyst

concentration

5 g/dL Zn+2 in THF

density

0.949 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Application

Rieke® Zinc metal can be used to reduce common organic functional groups such as aromatic nitro, conjugated aldehydes, arylacetylenes, and phenylpropiolates.

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.0 °F - closed cup

Flash Point(C)

-17.22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rieke zinc as a reducing agent for common organic functional groups
Kroemer J, et al.
Tetrahedron Letters, 47(36), 6339-6341 (2006)
M E Kuyumcu et al.
European review for medical and pharmacological sciences, 17(11), 1467-1471 (2013-06-19)
Zinc is one of the most important elements for human body. Zinc deficiency can occur in any age, if it is seen in elderly its clinical results can be more harmful due to already diminished functions. Some studies showed zinc
A viewpoint about the treatment of Wilson's disease.
Abdul Qayyum Rana et al.
The Canadian journal of neurological sciences. Le journal canadien des sciences neurologiques, 40(4), 612-614 (2013-06-22)
Marco Ranocchiari et al.
Chimia, 67(6), 397-402 (2013-08-16)
Structural features, synthesis, and reactivity of Zn-biphenyl metal-organic frameworks with MOF-5 topology are presented to show the chemical flexibility of such materials and to demonstrate the challenges that can be encountered and solved to avoid interpenetration. We introduce the synthesis
Impact of cadmium and zinc on root system of Lolium perenne and Trifolium repens.
T Lambrechts et al.
Communications in agricultural and applied biological sciences, 78(1), 19-24 (2013-07-24)

Articles

Metal-enhanced organic synthesis via organometallic intermediates is a widely used preparative route for thousands of organic compounds.

Metal-enhanced organic synthesis via organometallic intermediates is a widely used preparative route for thousands of organic compounds.

Metal-enhanced organic synthesis via organometallic intermediates is a widely used preparative route for thousands of organic compounds.

Metal-enhanced organic synthesis via organometallic intermediates is a widely used preparative route for thousands of organic compounds.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service