Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
Linear Formula:
CD3(CH2)4CO2H
CAS Number:
Molecular Weight:
119.18
NACRES:
NA.12
PubChem Substance ID:
UNSPSC Code:
12352106
MDL number:
Isotopic purity:
99 atom % D
Assay:
99% (CP)
Mass shift:
M+3
InChI
1S/C6H12O2/c1-2-3-4-5-6(7)8/h2-5H2,1H3,(H,7,8)/i1D3
SMILES string
[2H]C([2H])([2H])CCCCC(O)=O
InChI key
FUZZWVXGSFPDMH-FIBGUPNXSA-N
isotopic purity
99 atom % D
assay
99% (CP)
refractive index
n20/D 1.412 (lit.)
bp
202-203 °C (lit.)
mp
-3 °C (lit.)
density
0.951 g/mL at 25 °C
mass shift
M+3
Quality Level
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1C
Storage Class
8A - Combustible corrosive hazardous materials
wgk
WGK 1
flash_point_f
215.6 °F - closed cup
flash_point_c
102 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jingwei Cai et al.
Journal of proteome research, 15(2), 563-571 (2015-12-24)
Recent studies have identified the important role of the gut microbiota in the pathogenesis and progression of obesity and related metabolic disorders. The antioxidant tempol was shown to prevent or reduce weight gain and modulate the gut microbiota community in
Matthew V Gomez et al.
Toxicological sciences : an official journal of the Society of Toxicology, 179(1), 14-30 (2020-10-21)
The gut microbiome is a pivotal player in toxicological responses. We investigated the effects of maternal exposure to 3 human health-relevant toxicants (BDE-47, tetrabromobisphenol [TBBPA], and bisphenol S [BPS]) on the composition and metabolite levels (bile acids [BAs] and short-chain
D Nyasha Chagwedera et al.
Cell metabolism, 30(2), 364-373 (2019-05-28)
Microbial dysbiosis and inflammation are implicated in diet-induced obesity and insulin resistance. However, it is not known whether crosstalk between immunity and microbiota also regulates metabolic homeostasis in healthy animals. Here, we report that genetic deletion of tuberous sclerosis 1
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service