Skip to Content
Merck
All Photos(1)

Key Documents

489204

Sigma-Aldrich

Estrone-2,4,16,16-d4

95 atom % D

Synonym(s):

1,3,5(10)-Estratrien-3-ol-17-one-D4, 3-Hydroxy-1,3,5(10)-estratrien-17-one-D4, Deuterated estrone, Deuterated folliculin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18D4H18O2
CAS Number:
Molecular Weight:
274.39
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

95 atom % D

form

solid

optical activity

[α]20/D +161°, c = 1 in dioxane

technique(s)

mass spectrometry (MS): suitable

mp

258-260 °C (lit.)

mass shift

M+4

SMILES string

[2H]c1cc2[C@H]3CC[C@@]4(C)[C@@H](CC([2H])([2H])C4=O)[C@@H]3CCc2c([2H])c1O

InChI

1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-16,19H,2,4,6-9H2,1H3/t14-,15-,16+,18+/m1/s1/i3D,7D2,10D

InChI key

DNXHEGUUPJUMQT-QSPUTOQOSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weiwei Yu et al.
Journal of hazardous materials, 396, 122657-122657 (2020-05-04)
17β-Estradiol (17β-E2) as a non-conventional pollutant with high damage, the effective removal of 17β-E2 had been studied wildly. In recent years, nano materials application enabled the rapid removal of 17β-E2. Nano zero valent iron (nZVI) as one of the most
Stefan Poschner et al.
Frontiers in pharmacology, 8, 699-699 (2017-10-21)
The beneficial effect of dietary soy food intake, especially for women diagnosed with breast cancer, is controversial, as
Wei Chen et al.
Water research, 137, 211-219 (2018-03-20)
A passive water sampler based on the diffusive gradients in thin-films (DGT) technique was developed and tested for 3 groups of endocrine disrupting chemicals (EDCs, including oestrogens, alkyl-phenols and bisphenols). Three different resins (hydrophilic-lipophilic-balanced (HLB), XAD18 and Strata-XL-A (SXLA)) were
T Coello-Garcia et al.
Water research, 164, 114905-114905 (2019-08-09)
The removal of β-estradiol (E2) and α-ethinylestradiol (EE2) in biological wastewater treatment plants (WWTP) would need to be improved in order to comply with prospective Environmental Quality Standards (EQS) of 0.4 and 0.035 ng.L-1 respectively. The effluent concentration of a micropollutant
Xiaoguang Sunny Li et al.
Talanta, 178, 464-472 (2017-11-16)
Accurate quantitation of estrogens (i.e, estrone (E1), estradiol (E2) and estriol (E3)) is valuable for clinical assessment of human health and disease. Alterations in estrogen levels have been implicated in numerous pathological conditions. However, inadequacies in sensitivity and specificity, cumbersome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service