Skip to Content
Merck
All Photos(2)

Documents

482951

Sigma-Aldrich

Fullerene-C76

greener alternative

98%

Synonym(s):

Fullerene-C 76

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C76
CAS Number:
Molecular Weight:
912.81
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

98%

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>350 °C (lit.)

solubility

organic solvents: soluble

greener alternative category

General description

C76 is a higher fullerene with 76 carbon atoms. It may be prepared by dc arc discharge method. C NMR study of C76 shows the phase transition temperature to be in the range of 135-170K and the effect of crystallinity is different from C60 and C70. Electron affinity of C76 was determined to be 2.88+/- 0.05 eV.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Fullerene-C76 is a higher order fullerene with good optical absorption, which can be potentially used for polymeric solar cells based applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electron affinities of higher fullerenes
Boltalina OV, et al.
Rapid Communications in Mass Spectrometry, 7(11), 1009-1011 (1993)
Higher fullerenes as electron acceptors for polymer solar cells: A quantum chemical study
Morvillo P
Solar Energy Materials and Solar Cells, 93(10), 1827-1832 (2009)
Solid C76: NMR studies of molecular dynamics
Maniwa Y and Kume K
Physical Review, 53(21) (1996)
Hybrid silicon-carbon nanostructures for broadband optical absorption
Yang W, et al.
Royal Society of Chemistry Advances, 7(13), 8070-8076 (2017)
Selective and High Yield Synthesis of Higher Fullerenes
Tohji K, et al.
The Journal of Physical Chemistry, 99, 17785-17788 (1999)

Articles

Carbon nanotubes are materials that possess remarkable properties and offer extraordinary possibilities.

Find various photovoltaic and bioscience-based applications of fullerenes.

Review on 1D vdWHs: Discusses materials, synthesis, optoelectronic applications, challenges, and future perspectives for 1D vdWH-based devices.

Review on 1D vdWHs: Discusses materials, synthesis, optoelectronic applications, challenges, and future perspectives for 1D vdWH-based devices.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service