Skip to Content
Merck

482005

Poly(dimethylsiloxane), hydroxy terminated

viscosity ~50,000 cSt 

Synonym(s):

PDMS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

refractive index

n20/D 1.406

viscosity

~50,000 cSt

density

0.98 g/mL at 25 °C

SMILES string

C[Si](C)(O)O

InChI

1S/C2H8O2Si/c1-5(2,3)4/h3-4H,1-2H3

InChI key

XCLIHDJZGPCUBT-UHFFFAOYSA-N



Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library