Skip to Content
Merck
All Photos(2)

Key Documents

480061

Sigma-Aldrich

2,6-Dimethylphenylboronic acid

≥95.0%

Synonym(s):

2,6-Dimethylbenzeneboronic acid, 2,6-Xyleneboronic acid, 2,6-Xylylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3B(OH)2
CAS Number:
Molecular Weight:
149.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

impurities

<10% water

mp

105 °C (dec.) (lit.)

SMILES string

Cc1cccc(C)c1B(O)O

InChI

1S/C8H11BO2/c1-6-4-3-5-7(2)8(6)9(10)11/h3-5,10-11H,1-2H3

InChI key

ZXDTWWZIHJEZOG-UHFFFAOYSA-N

Application

Reagent used for
  • Palladium catalyzed Suzuki-Miyaura coupling reactions
  • One-pot ipso-nitration of arylboronic acids including broader substrate scope of heterocycles and functional groups
  • Nickel-Catalyzed Cross-Coupling of Chromene Acetals and Boronic Acids
  • Visible-light initiated aerobic oxidative hydroxylation catalyzed by Ru-complex
  • Rhodium(I)-catalyzed 1,4-addition reactions
  • Pd-catalyzed homocouplings
  • Expanded scope of Cu assisted Suzuki-Miyaura coupling reactions including aryl chlorides and polyhalo aryl boronates

Reagent used in Prepration of
  • Orally bioavialable G Protein-Coupled Receptor 40 agonists for diabetes treatment
  • Solid phase synthesis and antitumor structure-activity relationship of Smac triazoloprolines and biarylalanines tetrapeptide libraries
  • Protein Kinase inhibitors

Other Notes

contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sebastian T Le Quement et al.
ACS combinatorial science, 13(6), 667-675 (2011-09-13)
Apoptotic induction mechanisms are of crucial importance for the general homeostasis of multicellular organisms. In cancer the apoptotic pathways are downregulated, which, at least partly, is due to an abundance of inhibitors of apoptosis proteins (IAPs) that block the apoptotic
Thomas J A Graham et al.
Organic letters, 14(6), 1616-1619 (2012-03-06)
A modular and highly efficient protocol for the synthesis of 2-aryl- and heteroaryl-2H-chromenes is described. Under base-free conditions, readily accessible 2-ethoxy-2H-chromenes undergo C(sp(3))-O activation and C(sp(3))-C bond formation in the presence of an inexpensive nickel catalyst and boronic acids. This
Santos-Filho, E. F.; et al.
Tetrahedron Letters, 52, 5288-5288 (2011)
Room-temperature synthesis of tetra-ortho-substituted biaryls by NHC-catalyzed Suzuki-Miyaura couplings.
Linglin Wu et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(46), 12886-12890 (2011-10-11)
Satoshi Mikami et al.
Journal of medicinal chemistry, 55(8), 3756-3776 (2012-03-21)
As part of a program to identify potent GPR40 agonists with drug-like properties suitable for clinical development, the incorporation of polar substituents was explored with the intention of decreasing the lipophilicity of our recently disclosed phenylpropanoic acid derivative 1. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service