Skip to Content
Merck
All Photos(1)

Documents

478717

Sigma-Aldrich

2-(2-Nitrovinyl)furan

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO3
CAS Number:
Molecular Weight:
139.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

72-75 °C (lit.)

SMILES string

[O-][N+](=O)\C=C\c1ccco1

InChI

1S/C6H5NO3/c8-7(9)4-3-6-2-1-5-10-6/h1-5H/b4-3+

InChI key

WVUICGOYGDHVBH-ONEGZZNKSA-N

General description

2-(2-Nitrovinyl)furan (NVF) is a 2-furylethylene derivative with a potential antiprotozoal property. It forms solid inclusion complexes with cyclodextrine (CD) derivatives, 2-hydroxypropyl-β-cyclodextrin and sulfobutyl ether-β-cyclodextrin. The Friedel-Crafts alkylation of naphthols with NVF in aqueous medium and catalyst-free condition has been reported. The formation of Morita-Baylis-Hillman (MBH) adduct via MBH reaction of NVF with diisopropyl azodicarboxylate (DIAD) has been studied.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalyst-free Friedel-Crafts alkylation of naphthols with nitrostyrenes in the presence of water.
Halimehjani AZ, et al.
Tetrahedron Letters, 50(13), 1441-1443 (2009)
Diana Oliveira et al.
International journal of molecular sciences, 22(2) (2021-01-14)
Quorum sensing (QS) plays an essential role in the production of virulence factors, in biofilm formation and antimicrobial resistance. Consequently, inhibiting QS is being considered a promising target for antipathogenic/anti-virulence therapies. This study aims to screen 2-nitrovinylfuran derivatives structurally related
Characterization and molecular modeling of the inclusion complexes of 2-(2-nitrovinyl) furan (G-0) with cyclodextrines.
Ruz V, et al.
International Journal of Pharmaceutics, 439(1), 275-285 (2012)
T O Ajiboye et al.
Microbial pathogenesis, 91, 107-114 (2015-12-02)
The involvement of reactive oxygen species and oxidative stress in 2-(2-nitrovinyl) furan mediated bacterial cell death was investigated in Escherichia coli, Pseudomonas aeruginosa and Staphylococcus aureus. Time kill assay resulted in significant decrease in the optical density and colony-forming unit
Highly efficient hydrazination of conjugated nitroalkenes via imidazole or DMAP mediated Morita-Baylis-Hillman reaction.
Dadwal M, et al.
Organic & Biomolecular Chemistry, 4(13), 2525-2528 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service