Skip to Content
Merck
All Photos(2)

Key Documents

473340

Sigma-Aldrich

1,3,5-Triacryloylhexahydro-1,3,5-triazine

98%

Synonym(s):

1,3,5-Triacryloyl-s -triazine, 1,3,5-Triacryloylhexahydro-s -triazine, 1,3,5-Triacryloylhexahydrotriazine, Tri(N -acryloyl)hexahydrotriazine, Triacrylformal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N3O3
CAS Number:
Molecular Weight:
249.27
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

mp

156 °C (dec.) (lit.)

SMILES string

C=CC(=O)N1CN(CN(C1)C(=O)C=C)C(=O)C=C

InChI

1S/C12H15N3O3/c1-4-10(16)13-7-14(11(17)5-2)9-15(8-13)12(18)6-3/h4-6H,1-3,7-9H2

InChI key

FYBFGAFWCBMEDG-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Sereĭkaĭte et al.
Bioorganicheskaia khimiia, 29(3), 254-257 (2003-07-09)
The kinetics of the nucleophilic addition reactions of divinyl sulfone to amino groups of glycine and model proteins was studied in aqueous solution at 30 degrees C. The rate constants for glycine, bovine serum albumin, and alpha 1-casein were (4.84
G Dienys et al.
Bioconjugate chemistry, 11(5), 646-651 (2000-09-20)
Zn(2+) and Co(2+) ions are known to promote human growth hormone reversible dimerization. In these studies, dimerization was also shown to be initiated by nine other metal ions: Cd(2+), Hg(2+), Cu(2+), Ag+, Au(3+), Au+, Pd(2+), Ni(2+), and Pt(4+). In some
G Dienys et al.
Bioconjugate chemistry, 9(6), 744-748 (1998-11-17)
Six difunctional and trifunctional derivatives of acrylamide were synthesized and investigated as potential protein lysine residue cross-linking agents. 1,3,5-Triacryloyl-hexahydro-s-triazine (TAT) was considered the best. The rate constants for the reactions of TAT with model nucleophiles in water solution at 25
R S Slesinski et al.
Toxicology, 40(2), 145-163 (1986-08-01)
TAHT (1,3,5-triacryloylhexahydro-s-triazine), a reactive chemical coupling agent, was highly toxic following a single peroral dose of an aqueous suspension (10% w/v) to Wistar rats, or following application of TAHT in dichloromethane (DCM) solution (10% w/v) to covered skin of New
G Patras et al.
Electrophoresis, 21(17), 3843-3850 (2001-03-29)
The properties of polyacrylamide hydrogels synthesized with a novel hexafunctional (three double bonds) cross-linker, hexahydro-1,3,5-triacryloyl-s-triazine (1a), was evaluated and compared to the currently used tetrafunctional (two double bonds) cross-linker N,N-methylenebisacrylamide (Bis). A variety of characterization techniques that require very little

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service