471917
3,4-Dichlorophenylboronic acid
≥95%
Synonym(s):
3,4-Dichlorobenzeneboronic acid
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Quality Level
Assay
≥95%
form
powder
mp
280-285 °C (lit.)
SMILES string
OB(O)c1ccc(Cl)c(Cl)c1
InChI
1S/C6H5BCl2O2/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3,10-11H
InChI key
JKIGHOARKAIPJI-UHFFFAOYSA-N
Application
Reactant involved in:
Precursor / reactant involved in synthesis of biologically active molecules including:
- Lithiation/borylation-protodeboronation of homoallyl carbamates
- Suzuki coupling reactions
Precursor / reactant involved in synthesis of biologically active molecules including:
- Mycobacterium tuberculosis H37Rv chorismate mutase inhibitors
- Pyrrole derivatives as PDE4B inhibitors
- Nitrovinyl biphenyls as anticancer agents
- Dual immunosuppressive and anti-inflammatory agents
- Pyrazolopyrimidines as Cryptosporidium and Toxoplasma CDPK1 inhibitors
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service