Skip to Content
Merck
All Photos(2)

Key Documents

471291

Sigma-Aldrich

Isopropylamine

≥99.5%

Synonym(s):

2-Aminopropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNH2
CAS Number:
Molecular Weight:
59.11
Beilstein:
605259
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.04 (vs air)

Quality Level

vapor pressure

9.2 psi ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

755 °F

expl. lim.

10.4 %

impurities

≤0.1% water

color

APHA: ≤50

refractive index

n20/D 1.374 (lit.)

bp

33-34 °C (lit.)

density

0.688 g/mL at 20 °C (lit.)

functional group

amine

storage temp.

2-8°C

SMILES string

CC(C)N

InChI

1S/C3H9N/c1-3(2)4/h3H,4H2,1-2H3

InChI key

JJWLVOIRVHMVIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isopropylamine (i-PrNH2), a primary mine, is a urinary metabolite of propranolol.

Application

Isopropylamine was used in the following studies:
  • Preparation of enantiomerically pure (R and S)- acebutolol.
  • As template during the hydrothermal synthesis of SAPO materials.
  • Synthesis of bis(salicylaldiminate)nickel(II) complexes.
  • Preparation of hydrated, microporous aluminum phosphate AlPO-14.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

<-13.0 °F - closed cup

Flash Point(C)

<= -25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ethylene oligomerization by novel catalysts based on bis (salicylaldiminate) nickel (II) complexes and organoaluminum co-catalysts.
Carlini C, et al.
Applied Catalysis A: General, 231(1), 307-320 (2002)
On the possibility of incorporating Mn (II) and Cr (III) in SAPO-34 in the presence of isopropylamine as a template.
Rajic N, et al.
Zeolites, 13(4), 384-387 (1993)
Isopropylamine, a biologically active deamination product of propranolol in dogs: identification of deuterated and unlabeled isopropylamine by gas chromatography-mass spectrometry.
T Walle et al.
The Journal of pharmacology and experimental therapeutics, 183(3), 508-512 (1972-12-01)
Valter Castelvetro et al.
Journal of hazardous materials, 407, 124364-124364 (2020-11-04)
A novel procedure for nylon 6 and nylon 6,6 polyamide (PAs) microplastics (MPs) quantification is described for the first time. The overall procedure, including quantification of poly(ethylene terephthalate) (PET), was tested on wastewater treatment plant (WWTP) sludges. The three polymers
Huaixin Yang et al.
The journal of physical chemistry. B, 109(10), 4464-4469 (2006-07-21)
Differential scanning calorimetry of the hydrated, microporous aluminum phosphate AlPO-14 shows two distinct water losses between room temperature and 120 degrees C, indicating the presence of two types of water in the solid. Multiple-quantum magic angle spinning (MQMAS) (27)Al NMR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service