Skip to Content
Merck
All Photos(1)

Documents

469920

Sigma-Aldrich

2-Ethynylpyridine

98%

Synonym(s):

2-Pyridinylethyne, 2-Pyridylacetylene, Pyridin-2-ylacetylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5N
CAS Number:
Molecular Weight:
103.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.56 (lit.)

bp

85 °C/12 mmHg (lit.)

density

1.021 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C#Cc1ccccn1

InChI

1S/C7H5N/c1-2-7-5-3-4-6-8-7/h1,3-6H

InChI key

NHUBNHMFXQNNMV-UHFFFAOYSA-N

General description

2-Ethynylpyridine is a pyridine derivative.

Application

2-Ethynylpyridine has been used to synthesize:
  • poly(2-ethynyl-N-iodopyridinium iodide), via in-situ uncatalyzed polymerization
  • poly[2-ethynyl-N-(2-furoyl)pyridinium chloride
  • poly[2-ethynyl-N-(propylsulfonate)pyridinium betaine]
  • 4-(2′-pyridyl)-1,2,3-triphospholide anion, via reaction with anionic heptaphosphide clusters
  • poly[N-(carboxymethyl)-2-ethynylpyridinium bromide], a new water-soluble ionic conjugated polymer
  • self-doped ionic conjugated polymer, poly(2-ethynylpyridinimum N-benzoylsulfonate).

Other Notes

It may darken in storage with no loss in purity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert S P Turbervill et al.
Inorganic chemistry, 52(9), 5527-5534 (2013-04-24)
Reactions between anionic heptaphosphide clusters ([P7](3-)/[HP7](2-)) and 2-ethynylpyridine yielded the 4-(2'-pyridyl)-1,2,3-triphospholide anion ([P3C2H(2-C5H4N)](-); 1). This species was isolated as a compositionally pure [K(2,2,2-crypt)](+) salt in moderate yields. Preliminary coordination studies of 1 toward Mo(CO)6 or Mo(py)3(CO)3 (py = pyridine) afforded
Cecilia Zavala-Tecuapetla et al.
Pharmacological reports : PR, 66(5), 927-930 (2014-08-26)
Action of an antagonist of metabotropic glutamate receptors subtype 5 MTEP was studied in a model of complex partial seizures. Dorsal hippocampus of rat pups 12, 18 and 25 days old was stimulated six times with 10-min intervals. MTEP (20
Yeong-Soon Gal et al.
Journal of nanoscience and nanotechnology, 14(8), 6247-6250 (2015-05-06)
A new ionic polyacetylene derivative with furoyl substituents was prepared by the uncatalyzed polymerization of 2-ethynylpyridine by using 2-furoyl chloride in high yield. The polymer structure was characterized by such instrumental methods as NMR, IR, and UV-visible spectroscopies to have
Yeong-Soon Gal et al.
Journal of nanoscience and nanotechnology, 12(5), 4361-4364 (2012-08-03)
A new water-soluble ionic conjugated polymer, poly[N-(carboxymethyl)-2-ethynylpyridinium bromide], was prepared by the activated polymerization of 2-ethynylpyridine by using bromoacetic acid. This polymerization proceeded well in mild reaction conditions without any additional initiator or catalyst. The polymer structure was characterized by
Yeong-Soon Gal et al.
Journal of nanoscience and nanotechnology, 14(10), 8028-8032 (2015-05-07)
Poly(2-ethynyl-N-iodopyridinium iodide) [PEIPI] was easily prepared via in-situ uncatalyzed polymerization of 2-ethynylpyridine by using iodine. The activated acetylenic bond of 2-ethynyl-N-iodopyridinium iodide formed at the initial reaction time was assumed to be susceptible to linear polymerization, followed by an identical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service