Skip to Content
Merck
All Photos(1)

Key Documents

469165

Sigma-Aldrich

N-Z-L-serine methyl ester

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(NHCO2CH2C6H5)CO2CH3
CAS Number:
Molecular Weight:
253.25
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

reaction suitability

reaction type: solution phase peptide synthesis

bp

170 °C/0.01 mmHg (lit.)

mp

41-43 °C (lit.)

application(s)

peptide synthesis

SMILES string

COC(=O)[C@H](CO)NC(=O)OCc1ccccc1

InChI

1S/C12H15NO5/c1-17-11(15)10(7-14)13-12(16)18-8-9-5-3-2-4-6-9/h2-6,10,14H,7-8H2,1H3,(H,13,16)/t10-/m0/s1

InChI key

CINAUOAOVQPWIB-JTQLQIEISA-N

Application

This protected serine has been employed as a starting material for the Cbz analog of Garner′s aldehyde, 2,3-diaminopropanol, and optically active derivatives of 2-amino-1,3-propanediol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kang, M. et al.
The Journal of Organic Chemistry, 61, 5528-5528 (1996)
Demirci, F. et al.
Synthesis, 189-189 (1996)
Monache, G.D. et al.
Synthesis, 1155-1155 (1995)
James A. Marshall et al.
The Journal of organic chemistry, 61(2), 581-586 (1996-01-26)
The aminoheptose destomic acid (3.5) and the aminooctose lincosamine (6.8) were synthesized in protected form by parallel sequences starting from the oxazolidine derivatives 2.4 and 5.1 of N-CBz serinal and N-BOC threoninal. The parallel sequences feature BF(3)-promoted addition of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service