460044
5-Norbornene-2-endo,3-endo-dimethanol
98%
Synonym(s):
NDM
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H14O2
CAS Number:
Molecular Weight:
154.21
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
98%
bp
170-175 °C/25 mmHg (lit.)
mp
82-86 °C (lit.)
SMILES string
OC[C@@H]1[C@H]2C[C@H](C=C2)[C@@H]1CO
InChI
1S/C9H14O2/c10-4-8-6-1-2-7(3-6)9(8)5-11/h1-2,6-11H,3-5H2/t6-,7+,8-,9+
InChI key
IGHHPVIMEQGKNE-SPJNRGJMSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Brian D Johnston et al.
Antimicrobial agents and chemotherapy, 64(10) (2020-07-29)
Emerging carbapenem resistance in Escherichia coli, including sequence type 131 (ST131), the leading cause of extraintestinal E. coli infections globally, threatens therapeutic efficacy. Accordingly, we determined broth microdilution MICs for three distinctive newer agents, i.e., cefiderocol (CFDC), ceftazidime-avibactam (CZA), and
Miao He et al.
Acta diabetologica, 57(4), 479-489 (2019-11-22)
To compare circular (circRNA) expression levels in the vitreous humour between PDR (proliferative diabetic retinopathy) and the control groups. The present study collected vitreous humour samples of both the PDR group and the control group (composed of rhegmatogenous retinal detachment
Shamsi Khalid et al.
Microbial drug resistance (Larchmont, N.Y.), 26(3), 284-289 (2019-08-10)
The emergence of blaNDM particularly in Gram-negative bacteria is a burden on the health care system in developing countries. Hence, this study was initiated to screen New Delhi Metallo-β-lactamase (NDM)-producing Gram-negative bacterial strains from neonatal intensive care unit (NICU) of
Qian Ren et al.
Journal of diabetes and its complications, 34(2), 107464-107464 (2019-11-28)
Our aim was to search for clinical predictors of good glycemic control in patients starting or intensifying oral hypoglycemic pharmacological therapy. A multicenter, prospective cohort of 499 diabetic subjects was enrolled in this study: patients with newly diagnosed diabetes (NDM
Hyo Jeong Lee et al.
Pharmaceuticals (Basel, Switzerland), 14(1) (2021-01-10)
Rearranged during transfection (RET), a receptor tyrosine kinase, is activated by glial cell line-derived neurotrophic factor family ligands. Chromosomal rearrangement or point mutations in RET are observed in patients with papillary thyroid and medullary thyroid carcinomas. Oncogenic alteration of RET
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service