Skip to Content
Merck
All Photos(1)

Key Documents

457957

Sigma-Aldrich

Holmium

chips, 99.9% trace metals basis

Synonym(s):

Holmium-165

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ho
CAS Number:
Molecular Weight:
164.93
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

chips

reaction suitability

reagent type: catalyst
core: holmium

density

8.8 g/mL at 25 °C (lit.)

SMILES string

[Ho]

InChI

1S/Ho

InChI key

KJZYNXUDTRRSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerrit H van de Maat et al.
European radiology, 23(3), 827-835 (2012-09-28)
To demonstrate the feasibility of MRI-based assessment of the intrahepatic Ho-PLLA-MS biodistribution after radioembolisation in order to estimate the absorbed radiation dose. Fifteen patients were treated with holmium-166 ((166)Ho) poly(L-lactic acid)-loaded microspheres (Ho-PLLA-MS, mean 484 mg; range 408-593 mg) in
Alexander Hemming et al.
Optics express, 21(4), 4560-4566 (2013-03-14)
We report the highest power operation of a resonantly cladding-pumped, holmium-doped silica fibre laser. The cladding pumped all-glass fibre utilises a fluorine doped glass layer to provide low loss cladding guidance of the 1.95 µm pump radiation. The operation of
Peter R Seevinck et al.
International journal of radiation oncology, biology, physics, 83(3), e437-e444 (2012-05-29)
To investigate the potential of magnetic resonance imaging (MRI) for accurate assessment of the three-dimensional (166)Ho activity distribution to estimate radiation-absorbed dose distributions in (166)Ho-loaded poly (L-lactic acid) microsphere ((166)Ho-PLLA-MS) liver radioembolization. MRI, computed tomography (CT), and single photon emission
Maarten L J Smits et al.
The Lancet. Oncology, 13(10), 1025-1034 (2012-08-28)
The efficacy of radioembolisation for the treatment of liver tumours depends on the selective distribution of radioactive microspheres to tumorous tissue. The distribution of holmium-166 ((166)Ho) poly(L-lactic acid) microspheres can be visualised in vivo by both single-photon-emission CT (SPECT) and
Y Nedjadi et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 70(9), 1990-1996 (2012-03-17)
The work presented here is a new and precise measurement of the half-life of (166m)Ho by determining the activity concentration, using an ionisation chamber calibrated for this nuclide, and measuring the number of (166m)Ho atoms using multi-collector inductively coupled plasma

Articles

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service