455652
Sodium p-toluenesulfinate
95%
Synonym(s):
p-Toluenesulfinic acid sodium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
4-CH3C6H4SO2Na
CAS Number:
Molecular Weight:
178.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95%
impurities
<5% water
mp
>300 °C (lit.)
functional group
sulfinic acid
SMILES string
[Na+].Cc1ccc(cc1)S([O-])=O
InChI
1S/C7H8O2S.Na/c1-6-2-4-7(5-3-6)10(8)9;/h2-5H,1H3,(H,8,9);/q;+1/p-1
InChI key
KFZUDNZQQCWGKF-UHFFFAOYSA-M
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis of cyanohydrin trimethylsilyl ethers catalyzed by potassium p-toluenesulfinate.
Dekamin MG, et al.
Catalysis Communications, 9(6), 1352-1355 (2008)
Sodium p-Toluenesulfinate in Free Radical Reactions.
Chuang C-P.
Synthetic Communications, 23(17), 2371-2380 (1993)
Sodium p-toluenesulfinate/copper (II) acetate in free radical reactions of 5-aryl substituted alkenes.
Wang S-F, et al.
Tetrahedron, 55(8), 2273-2288 (1999)
Efficient and Selective Trimerization of Aryl and Alkyl Isocyanates Catalyzed by Sodium p-Toluenesulfinate in the Presence of TBAI in a Solvent-Free Condition.
Moghaddam FM, et al.
Bulletin of the Chemical Society of Japan, 75(4), 851-852 (2002)
Asymmetric induction in the palladium-catalyzed sulfonylation of allylic sulfinates and acetates with chiral phosphine ligands.
Hiroi K and Makino K.
Chemistry Letters (Jpn), 15(4), 617-620 (1986)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service