Skip to Content
Merck
All Photos(3)

Key Documents

451622

Sigma-Aldrich

Lithium tetrafluoroborate

greener alternative

ultra dry, powder, 99.99% trace metals basis

Synonym(s):

Lithium borofluoride, Lithium fluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiBF4
CAS Number:
Molecular Weight:
93.75
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
form:
powder

description

Grade: ultra dry

Quality Level

Assay

99.99% trace metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤150.0 ppm Trace Metal Analysis

mp

293-300 °C (dec.) (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

[Li+].F[B-](F)(F)F

InChI

1S/BF4.Li/c2-1(3,4)5;/q-1;+1

InChI key

UFXJWFBILHTTET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium tetrafluoroborate is a class of electrolytic materials that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

LiBF4 is majorly used as an electrolyte with good ionic conductivity. It facilitates the formation of an electrolytic solution or a liquid electrolyte, which can be used in the fabrication of lithium-ion batteries and solid-state electrochemical displays.
Used recently in the development of a solid composite electrolyte for lithium batteries.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of di-ureasil ormolytes based on lithium tetrafluoroborate in solid-state electrochromic displays
Barbosa P, et al.
Journal of Materials Chemistry, 20(4), 723-730 (2010)
A new lithium salt with dihydroxybenzene and lithium tetrafluoroborate for lithium battery electrolytes
Xue Z, et al.
Journal of Power Sources, 196(20), 8710-8713 (2011)
Zachary T Gossage et al.
The Analyst, 145(7), 2631-2638 (2020-02-27)
Interphases formed at battery electrodes are key to enabling energy dense charge storage by acting as protection layers and gatekeeping ion flux into and out of the electrodes. However, our current understanding of these structures and how to control their
Proc. Power Sources Conf., 37th, 287-287 (1996)
Novel polymer electrolyte from poly (carbonate-ether) and lithium tetrafluoroborate for lithium-oxygen battery
Lu Q, et al.
Journal of Power Sources, 242, 677-682 (2013)

Articles

Nanomaterials for Energy Storage in Lithium-ion Battery Applications

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service