450464
4,4′-(1,3-Phenylenediisopropylidene)bisphenol
99%
Synonym(s):
Bisphenol M
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
C6H4[C(CH3)2C6H4OH]2
CAS Number:
Molecular Weight:
346.46
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
99%
mp
135-139 °C (lit.)
SMILES string
CC(C)(c1ccc(O)cc1)c2cccc(c2)C(C)(C)c3ccc(O)cc3
InChI
1S/C24H26O2/c1-23(2,17-8-12-21(25)13-9-17)19-6-5-7-20(16-19)24(3,4)18-10-14-22(26)15-11-18/h5-16,25-26H,1-4H3
InChI key
PVFQHGDIOXNKIC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Repr. 2 - Skin Sens. 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Martin Eckardt et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 37(10), 1791-1810 (2020-07-25)
Resoles are multifarious pre-polymeric resins produced by the condensation of basic chemicals phenols, formaldehyde and optionally aliphatic alcohols like butanol. They are widely used as cross-linkers to form resistant internal coatings on metal surfaces of cans, containers or closures. Although
Fiorella Lucarini et al.
International journal of environmental research and public health, 17(13) (2020-07-09)
Restrictions on the use of bisphenol A (BPA) in consumer products led to its replacement by various bisphenol (BP) analogues, yet young children's exposure to these analogues has been poorly characterized so far. This study aimed to characterize infants' and
Na Li et al.
Environmental pollution (Barking, Essex : 1987), 271, 116394-116394 (2021-01-04)
Common plasticizers and their alternatives are environmentally ubiquitous and have become a global problem. In this study, common plasticizers (phthalates and metabolites) and new alternatives [bisphenol analogs, t-butylphenyl diphenyl phosphate (BPDP), and bisphenol A bis(diphenyl phosphate) (BDP)] were quantified in
Stefan Pj van Leeuwen et al.
Chemosphere, 221, 246-253 (2019-01-15)
Information on the occurrence and endocrine potencies of analogues of bisphenol A (BPA) and diglycidyl ester derivatives (BDGEs) of BPA and BPF is limited. Such information is, however, important as the current debate on BPA and the lowered BPA migration
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service