Skip to Content
Merck
All Photos(1)

Documents

445568

Sigma-Aldrich

Calcium methoxide

97%

Synonym(s):

Calcium dimethoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(OCH3)2
CAS Number:
Molecular Weight:
102.15
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid
solid

reaction suitability

core: calcium

mp

>385 °C (lit.)

SMILES string

CO[Ca]OC

InChI

1S/2CH3O.Ca/c2*1-2;/h2*1H3;/q2*-1;+2

InChI key

AMJQWGIYCROUQF-UHFFFAOYSA-N

General description

Calcium methoxide is an alkaline earth metal compound that can be used as a calcium catalyst. It is a solid base catalyst that shows excellent catalytic activity and stability. It can be synthesized by reacting calcined calcium carbonate with methanol reflux under a nitrogen flow.

Application

Calcium methoxide can be used as a catalyst in the transesterification of oil to produce biodiesel.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transesterification of Nannochloropsis oculata microalga's oil to biodiesel using calcium methoxide catalyst
Teo SH, et al.
Energy, 78(7), 63-71 (2014)
Synthesis and kinetics of biodiesel formation via calcium methoxide base catalyzed transesterification reaction in the absence and presence of ultrasound
Deshmane, Vishwanath Ganpat and Adewuyi, Yusuf Gbadebo
Fuel: The Science and Technology of Fuel and Energy, 107(7), 474-482 (2013)
Calcium methoxide as a solid base catalyst for the transesterification of soybean oil to biodiesel with methanol
Liu X, et al.
Fuel: The Science and Technology of Fuel and Energy, 87(7), 1076-1082 (2008)
Rapeseed oil methyl esters preparation using heterogeneous catalysts
Gryglewicz S
Bioresource Technology, 70(3), 249-253 (1999)

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service