Skip to Content
Merck
All Photos(1)

Documents

443875

Sigma-Aldrich

Holmium(III) perchlorate solution

40 wt. % in H2O, 99.9% trace metals basis

Synonym(s):

Holmium perchlorate (Ho(ClO4 )3 ), Holmium triperchlorate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ho(ClO4)3
CAS Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

0.12 psi ( 20 °C)
1.68 psi ( 55 °C)

Quality Level

Assay

99.9% trace metals basis

form

liquid

reaction suitability

reagent type: catalyst
core: holmium

concentration

40 wt. % in H2O

impurities

≤1500.0 ppm Trace Rare Earth Analysis

density

1.57 g/mL at 25 °C

SMILES string

[Ho+3].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/3ClHO4.Ho/c3*2-1(3,4)5;/h3*(H,2,3,4,5);/q;;;+3/p-3

InChI key

HRWMCDDQLSZQPE-UHFFFAOYSA-K

General description

Holmium (III) perchlorate is a yellow solution widely used to verify the wavelength scale of UV/visible spectrophotometers with sharp absorption peaks at 200-900 nm.

Application

Holmium(III) perchlorate solution can be used:
  • As a precursor to synthesize Holmium(III) complex nanoparticles.
  • To study the stability of holmium oxide for the establishment of wavelength standards for spectrophotometer.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Liq. 2 - Skin Irrit. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Characterization of 18-and 20-Membered Dioxa Tetraaza Macrocylic Dysprosium (III) and Holmium (III) Complex Nanoparticles Encapsulated Within Nanopores of Zeolite Y
Maryam Shaterian, et al.
Synthesis and Reactivity in Inorganic, Metal-Organic, and Nano-Metal Chemistry, 46, 400-404 (2016)
High-resolution spectroscopy of Holmium Perchlorate: Establishment of wavelength standard for spectrophotometer
Anju, et al.
Optical Materials, 138, 113707-113707 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service