Skip to Content
Merck
All Photos(3)

Key Documents

439932

Sigma-Aldrich

4-Acetylphenyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COC6H4NCO
CAS Number:
Molecular Weight:
161.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

139 °C/14 mmHg (lit.)

mp

35-37 °C (lit.)

functional group

isocyanate
ketone

SMILES string

CC(=O)c1ccc(cc1)N=C=O

InChI

1S/C9H7NO2/c1-7(12)8-2-4-9(5-3-8)10-6-11/h2-5H,1H3

InChI key

MOZSVHZOUDIZMF-UHFFFAOYSA-N

General description

4-Acetylphenyl isocyanate is a reactive isocyanate and had one carbamate unit for every 7.6 graphene carbons.

Application

4-Acetylphenyl isocyanate may be used for the preparation of isocyanate-treated graphene oxide (iGO). It may be used for the following syntheses:
  • (4S,5R)-2-oxo-4-(4-phenyl-piperazin-1-ylmethyl)-oxazolidin-5-ylmethyl (4-acetyl-phenyl)-carbamate
  • (4S,5R)-2-oxo-4-(4-phenyl-piperazin-1-ylmethyl)-oxazolidin-5-ylmethyl (4-acetyl-phenyl)-carbamate
  • 2-oxo-4-(4-phenyl-piperazin-1-ylmethyl)-oxazolidin-5-ylmethyl phenyl acetate
  • 2-oxo-4-(4-phenyl-piperazin-1-ylmethyl)-oxazolidin-5-ylmethyl (4-acetyl-phenyl)-carbamate

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crina M Orac et al.
Journal of medicinal chemistry, 54(19), 6786-6795 (2011-08-05)
The enantiomers and the cis isomers of two previously studied 4,5-disubstituted oxazolidinones have been synthesized, and their binding to the T-box riboswitch antiterminator model RNA has been investigated in detail. Characterization of ligand affinities and binding site localization indicates that
Haiyang Zhao et al.
Physical chemistry chemical physics : PCCP, 15(23), 9084-9092 (2013-05-07)
In this paper, isocyanate-treated graphene oxide (iGO), which can be well dispersed in organic solvent, was prepared in a simple manner and showed excellent compatibility with polysulfone (PSF). iGO-PSF ultrafiltration membranes were prepared by the classical phase inversion method. The
Synthesis and exfoliation of isocyanate-treated graphene oxide nanoplatelets.
Stankovich S, et al.
Carbon, 44(15), 3342-3347 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service