Skip to Content
Merck
All Photos(2)

Key Documents

438960

Sigma-Aldrich

(R)-(+)-3-Benzyloxy-1,2-propanediol

99%

Synonym(s):

(R)-Glycerol 1-benzyl ether, 3-O-Benzyl-sn-glycerol, 3-Benzyl-sn-glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
182.22
Beilstein:
2048922
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

optical activity

[α]20/D +5.5°, c = 20 in chloroform

mp

25-29 °C (lit.)

storage temp.

2-8°C

SMILES string

OC[C@@H](O)COCc1ccccc1

InChI

1S/C10H14O3/c11-6-10(12)8-13-7-9-4-2-1-3-5-9/h1-5,10-12H,6-8H2/t10-/m1/s1

InChI key

LWCIBYRXSHRIAP-SNVBAGLBSA-N

Application

Employed in the synthesis of a cyclopropyl chiron used in the preparation of 2,3-methanoamino acids. Chiral building block in the synthesis of petrosynes and biologically important phospholipid analogs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burgess, K. et al.
The Journal of Organic Chemistry, 58, 3767-3767 (1993)
Iguchi, K. et al.
The Journal of Organic Chemistry, 58, 5690-5690 (1993)
S.F. Martin et al.
The Journal of Organic Chemistry, 59, 4805-4805 (1994)
T N Wheeler et al.
Journal of medicinal chemistry, 37(24), 4118-4129 (1994-11-25)
The substrate specificity at the active site of recombinant human synovial fluid phospholipase A2 (hs-PLA2) was investigated by the preparation of a series of short-chain phospholipid analogs and measurement of their enzymatic hydrolysis at concentrations well below the critical micelle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service