Skip to Content
Merck
All Photos(1)

Key Documents

436860

Sigma-Aldrich

N,N-Diisopropylaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N[CH(CH3)2]2
CAS Number:
Molecular Weight:
177.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.519 (lit.)

bp

95-96 °C/11 mmHg (lit.)

density

0.91 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CC(C)N(C(C)C)c1ccccc1

InChI

1S/C12H19N/c1-10(2)13(11(3)4)12-8-6-5-7-9-12/h5-11H,1-4H3

InChI key

OVSARSKQWCLSJT-UHFFFAOYSA-N

General description

N,N-Diisopropylaniline is an N,N-dialkylaniline. Its borane adducts have been prepared and reported as hydroborating agents. Thermal dissociation of the borane-N,N-diisopropylaniline adduct has been reported to afford gaseous diborane. N,N-Diisopropylaniline is a diisoproplyamino derivative that can be prepared by reacting bromobenzene with diisopropylamine.

Application

N,N-Diisopropylaniline was used for the synthesis of 4-diisopropylamino benzonitrile. It may be used for the synthesis of 6-(4-bromophenyl)-3-methoxy-5-methyl-8-oxabicyclo[3.2.1]octa-3,6-dien-2-one and N,N,N′,N′-tetraisopropylbenzidine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Synthesis of N,N,N',N'-tetraalkylbenzidines through oxidative coupling of N,N-dialkylarylamines induced by SbCl5.
Vitale P, et al.
ARKIVOC (Gainesville, FL, United States), 3, 36-48 (2013)
N-alkylation of hindered secondary aromatic amines with 2-iodobutane.
Katritzky AR, et al.
ORL; Journal for Oto-Rhino-Laryngology and Its Related Specialties, 23(4), 399-402 (1991)
Yvonne D Williams et al.
The Journal of organic chemistry, 78(23), 11707-11713 (2013-11-01)
Methoxytropolones are useful scaffolds for therapeutic development because of their known biological activity and established value in the synthesis of α-hydroxytropolones. Upon treatment with triflic acid, a series of 3-methoxy-8-oxabicyclo[3.2.1]octa-3,6-dien-2-ones rearrange rapidly and cleanly to form methoxytropolones. Interestingly, bicycles that
Brown et al.
The Journal of organic chemistry, 65(15), 4655-4661 (2000-08-26)
Several N,N-diethyl-tert-alkylamines, such as N,N-diethyl-2-methyl-2-butylamine (1, t-PentNEt2), N,N-diethyl-2,3-dimethyl-2-butylamine (2, t-HexNEt2), N,N-diethyl-2,3,3-trimethyl-2-butylamine (3, t-HeptNEt2), and N,N-diethyl-1,1,3,3-tetramethylbutylamine (4, t-OctNEt2) with varying steric bulk around nitrogen (by changing the tert-alkyl group) have been prepared and examined as borane carriers. The complexing ability of
Dual fluorescence and fast intramolecular charge transfer with 4-(diisopropylamino) benzonitrile in alkane solvents.
Demeter A, et al.
Chemical Physics Letters, 323(3), 351-360 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service