Skip to Content
Merck
All Photos(1)

Key Documents

436844

Sigma-Aldrich

3-Thienylboronic acid

≥95.0%

Synonym(s):

(Thiophene-3-yl)boronic acid, 3-Thiopheneboric acid, 3-Thiopheneboronic acid, 3-Thiophenylboronic acid, Thiophene-3-boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5BO2S
CAS Number:
Molecular Weight:
127.96
Beilstein:
113573
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥95.0%

form

solid

mp

164-169 °C (lit.)

SMILES string

OB(O)c1ccsc1

InChI

1S/C4H5BO2S/c6-5(7)4-1-2-8-3-4/h1-3,6-7H

InChI key

QNMBSXGYAQZCTN-UHFFFAOYSA-N

Other Notes

Contains varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Liang et al.
Dalton transactions (Cambridge, England : 2003), 49(10), 3326-3332 (2020-02-27)
The post-functionalization of six novel symmetric and asymmetric meso-expanded Co(iii) corroles through Suzuki-Miyaura coupling reactions has been successfully accomplished and is reported along with their structural characterization. An analysis of the structure-property relationships of the optical and redox properties of
Muamer Dervisevic et al.
Biosensors & bioelectronics, 90, 6-12 (2016-11-21)
The detection of cancer cells through important molecular recognition target such as sialic acid is significant for the clinical diagnosis and treatment. There are many electrochemical cytosensors developed for cancer cells detection but most of them have complicated fabrication processes
Hong-Min Meng et al.
The Analyst, 143(20), 4967-4973 (2018-09-19)
A multifunctional nanosystem, which integrates biosensing, bioimaging, and therapeutic functions into a single nanoprobe, is of great significance for biosensing and biomedicine. Near-infrared (NIR) graphene quantum dots (GQDs) have emerged as an attractive bioimaging and therapy tool for exploring biological
Muamer Dervisevic et al.
Materials science & engineering. C, Materials for biological applications, 72, 641-649 (2016-12-28)
In this study we report a new, simple and first impedimetric biosensor based on 3-Thienyl boronic acid for dopamine detection. Biosensor electrode preparation is 1min long by simple electro-polymerization of 3-Thienyl boronic acid and copolymer Thiophene P(TBA
Ingrid Benachio et al.
Journal of molecular recognition : JMR, 34(3), e2878-e2878 (2020-10-07)
Antibiotics, although being amazing compounds, need to be monitored in the environment and foodstuff. This is primarily to prevent the development of antibiotic resistance that may make them ineffective. Unsurprisingly, advances in analyticalsciences that can improve their determination are appreciated.

Articles

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service