Skip to Content
Merck

435546

Ethylenediamine tetrakis(ethoxylate-block-propoxylate) tetrol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
[-CH2N[(-CH2CH2O-)x[-CH2CH(CH3)O-]yH]2]2
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

suspension

Quality Segment

surface tension

43 dyn/cm, 25 °C, 0.1 wt. % in H2O

viscosity

3,870 cP(25 °C, Brookfield)(lit.)

mp

21 °C (DSC at peak)

transition temp

cloud point 71-75 °C

density

1.05 g/mL at 25 °C

HLB

1.0 - 7.0

SMILES string

N(CCN(CC(OCC(OCCO)C)C)CC(OCC(OCCO)C)C)(CC(OCC(OCCO)C)C)CC(OCC(OCCO)C)C

InChI

1S/C34H72N2O12/c1-27(45-23-31(5)41-15-11-37)19-35(20-28(2)46-24-32(6)42-16-12-38)9-10-36(21-29(3)47-25-33(7)43-17-13-39)22-30(4)48-26-34(8)44-18-14-40/h27-34,37-40H,9-26H2,1-8H3

InChI key

BFKFABWTAFNFID-UHFFFAOYSA-N

General description

Terminal secondary tetrahydroxy-functional oligomer.

Application

Hard and soft surface cleaners, defoamers in coatings and water treatment. Lubricant in metal working, anti-foaming aid and extender for linear and cross-linked polyesters and polyurethanes.

Features and Benefits

Ethylenediamine tetrakis(ethoxylate-block-propoxylate) tetrol exhibits surface activity that varies with the EO/PO ratio. It offers lower foaming properties, enhanced defoaming capabilities, and reduced gelling tendencies compared to typical surfactants. Additionally, its terminal secondary hydroxyl groups exhibit lower reactivity and acidity than primary hydroxyl groups found in other surfactants, making it a versatile choice for various applications.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1B

Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

>482.0 °F - closed cup

Flash Point (°C)

> 250 °C - closed cup

PPE (personal protective equipment)

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library