Skip to Content
Merck
All Photos(2)

Key Documents

432881

Sigma-Aldrich

4-(Chloromethyl)phenyl acetate

98%

Synonym(s):

α-Chloro-p-cresol acetate, 4-Acetoxybenzyl chloride, p-(Chloromethyl)phenyl acetate, p-Acetoxybenzyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C6H4CH2Cl
CAS Number:
Molecular Weight:
184.62
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.53 (lit.)

bp

240 °C (lit.)

density

1.201 g/mL at 25 °C (lit.)

SMILES string

CC(=O)Oc1ccc(CCl)cc1

InChI

1S/C9H9ClO2/c1-7(11)12-9-4-2-8(6-10)3-5-9/h2-5H,6H2,1H3

InChI key

OKRCXSCDWKJWSW-UHFFFAOYSA-N

Application

4-(Chloromethyl)phenyl acetate may be used as reagent in 4-(chloromethyl)phenyl acetate assay.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Maes et al.
Molecular neurobiology, 57(5), 2333-2345 (2020-02-11)
Accumulating evidence suggests that TNF-α-mediated immune-neurotoxicity contributes to cognitive impairments and the overall severity of schizophrenia (OSOS). There are no data whether peripheral IL-6 and IL-4 may affect the phenome of schizophrenia above and beyond the effects of TNF-α and
Michael Maes et al.
Molecular neurobiology, 56(9), 6626-6644 (2019-03-27)
Although, staging models gained momentum to stage define affective disorders, no attempts were made to construct mathematical staging models using clinical and biomarker data in patients with major depression and bipolar disorder. The aims of this study were to use
Michael Maes et al.
Metabolic brain disease, 36(3), 509-521 (2021-01-08)
Current diagnoses of mood disorders are not cross validated. The aim of the current paper is to explain how machine learning techniques can be used to a) construct a model which ensembles risk/resilience (R/R), adverse outcome pathways (AOPs), staging, and
Determination of paraoxonase 1 status without the use of toxic organophosphate substrates.
Rebecca J Richter et al.
Circulation. Cardiovascular genetics, 1(2), 147-152 (2009-12-25)
Dongyu Li et al.
Journal of agricultural and food chemistry, 65(20), 4209-4215 (2017-05-06)
A new near-infrared fluorescence off-on probe was developed and applied to fluorescence imaging of carboxylesterase in living HepG-2 cells and zebrafish pretreated with pesticides (carbamate, organophosphorus, and pyrethroid). The probe was readily prepared by connecting (4-acetoxybenzyl)oxy as a quenching and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service