Skip to Content
Merck
All Photos(2)

Key Documents

412910

Sigma-Aldrich

Methyltrioxorhenium(VII)

Re 71.0-76.0 %

Synonym(s):

MTO, Methylrhenium(VII) trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3ReO3
CAS Number:
Molecular Weight:
249.24
Beilstein:
3927932
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

composition

Re, 71.0-76.0%

reaction suitability

core: rhenium
reagent type: catalyst

mp

111 °C (lit.)

SMILES string

C[Re](=O)(=O)=O

InChI

1S/CH3.3O.Re/h1H3;;;;

InChI key

PQTLALPZRPFYIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Potent catalytic oxidant for converting alkenes to epoxides in a variety of solvents and 2-methylnaphthalene to 2-methyl-1,4-naphthoquinone (Vitamin K3).
Catalytic oxidant used with urea hydrogen peroxide in an efficient conversion of imines to nitrones.
An effective antioxidant under various conditions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven W Baldwin et al.
Organic letters, 6(10), 1653-1656 (2004-05-07)
Cycloaddition reactions between a new homochiral imidazolone-derived nitrone afford cycloadducts in high yield and with high stereoselectivity. Subsequent cycloadduct elaboration affords the gamma-lactones of gamma-hydroxy-alpha-amino acids as well as the optically pure amino acids themselves.
Inorganic Chemistry, 32, 4517-4517 (1993)
Veronika Huntosova et al.
Molecules (Basel, Switzerland), 26(2) (2021-01-23)
Detection of tissue and cell oxygenation is of high importance in fundamental biological and in many medical applications, particularly for monitoring dysfunction in the early stages of cancer. Measurements of the luminescence lifetimes of molecular probes offer a very promising
Bee Hong Soon et al.
Frontiers in physiology, 8, 231-231 (2017-05-10)
The role of mitochondria in tumorigenesis has regained much attention as it could dysregulate cellular energetics, oxidative stress and apoptosis. However, the role of mitochondria in different grade gliomasis still unknown. This study aimed to identify mitochondrial DNA (mtDNA) sequence
Applied Homogeneous Catalysis with Organometallic Compounds, 3, 1304-1304 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service