Skip to Content
Merck
All Photos(1)

Key Documents

412198

Sigma-Aldrich

Trimethylolpropane ethoxylate triacrylate

average Mn ~912

Synonym(s):

Trimethylolpropane ethoxylate (14/3 EO/OH) triacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=CHCO2(CH2CH2O)nCH2]3CC2H5
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

viscous liquid

Quality Level

mol wt

average Mn ~912

refractive index

n20/D 1.471 (lit.)

bp

157 °C (lit.)

density

1.12 g/mL at 25 °C

SMILES string

CCC(COCCOC(=O)C=C)(COCCOC(=O)C=C)COCCOC(=O)C=C

InChI

1S/C21H32O9/c1-5-18(22)28-12-9-25-15-21(8-4,16-26-10-13-29-19(23)6-2)17-27-11-14-30-20(24)7-3/h5-7H,1-3,8-17H2,4H3

InChI key

MTPIZGPBYCHTGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Robust and highly ion-conducting gel polymer electrolytes with semi-interpenetrating polymer network structure: Discusses the synthesis of polymer electrolytes using trimethylolpropane ethoxylate triacrylate (ETPTA) and their applications in ion-conducting materials (MG Gu et al., 2021).

  • Fabrication and characterization of thiol-triacrylate polymer via Michael addition reaction for biomedical applications: Explores the fabrication of thiol-triacrylate polymers using trimethylolpropane ethoxylate triacrylate for biomedical applications (A Forghani et al., 2018).

  • Preparation and Properties of UV Curable Hard Coating Materials on PET: Investigates the use of trimethylolpropane ethoxylate triacrylate (TMPETA) to improve the properties of UV-cured films on PET substrates (H Kim, 2014).

  • Photo-polymerized organic host network of ionogels for lithium batteries: effects of mesh size and of ethylene oxide content: Examines the effects of mesh size and ethylene oxide content in ionogels synthesized with trimethylolpropane ethoxylate triacrylate for lithium batteries (D Aidoud et al., 2018).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service