Skip to Content
Merck
All Photos(1)

Key Documents

40766

Sigma-Aldrich

N,N-Dimethylmethyleneiminium chloride

≥95.0% (AT)

Synonym(s):

Böhme′s salt, Böhme′s salt, Dimethylformiminium chloride, Methylenedimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=N+(CH3)2Cl-
CAS Number:
Molecular Weight:
93.56
Beilstein:
505955
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (AT)

form

powder

reaction suitability

reaction type: C-C Bond Formation

mp

146-148 °C (lit.)

functional group

amine

SMILES string

[Cl-].C[N+](C)=C

InChI

1S/C3H8N.ClH/c1-4(2)3;/h1H2,2-3H3;1H/q+1;/p-1

InChI key

ZJTROANVDZIEGB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Dimethylmethyleneiminium chloride (Böhme′s salt) is a dimethylaminomethylating agent that can be used as one of the key reagents in the following applications:
  • Electrophilic aminomethylation of aldehydes and ketones to synthesize corresponding Mannich products, also known as Mannich reaction.
  • Preparation of dihydronaphthyridinones as HIV-1 integrase inhibitors.
  • Preparation of cyanotetrahydrooxo-β-carbolines via cyclocondensation of cyanomethyl indole-2-carboxylate with ammonia.
  • Asymmetric synthesis of phalarine via stereospecific Pictet-Spengler cyclocondensation and traceless chirality transfer from L-tryptophan.
  • Synthesis of functionalized diarylpyrrolizines as inhibitors of microsomal prostaglandin E2 synthase-1 (mPGES-1) and 5-lipoxygenase (5-LOX).
  • Synthesis of (±)-phalarine with the rearrangement of an azaspiroindolenine and Gassman oxindole preparation as key steps.

Reacant for:
Preparation of dihydronaphthyridinones as HIV-1 integrase inhibitors
Mannich reactions
Preparation of cyanotetrahydrooxo-ß-carbolines via cyclocondensation reactions
Asymmetric synthesis of phalarine via stereospecific Pictet-Spengler cyclocondensation and traceless chirality transfer from L-tryptophan
Synthesis of functionalized diarylpyrrolizines as inhibitors of microsomal prostaglandin E2 synthase-1 (mPGES-1) and 5-lipoxygenase (5-LOX)
Synthesis of (±)-phalarine with the rearrangement of an azaspiroindolenine and Gassman oxindole preparation as key steps

Other Notes

"Mannich-reagent" for direct use; higher yields of purer products were achieved in shorter times compared with the classical "Mannich reaction"; in-situ preparation of the reactive triflate with TMS-triflate

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N. Holy et al.
Tetrahedron, 35, 613-613 (1979)
otal Synthesis of Phalarine.
Li C, et al.
Angewandte Chemie (International Edition in English), 46(9), 1448-1450 (2007)
H. Bohme
Angewandte Chemie (International Edition in English), 88, 772-772 (1976)
The Mannich reaction-II: derivatization of aldehydes and ketones using dimethyl (methylene) ammonium salts.
Holy N, et al.
Tetrahedron, 35(5), 613-619 (1979)
H.-U. Reissig et al.
Liebigs Ann. Chem., 1914-1914 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service