406686
Poly(propylene glycol) bis(2-aminopropyl ether)
average Mn ~2,000
Synonym(s):
Diaminopolypropylene glycol, Poly(oxypropylene)diamine, Polypropylene glycol diamine
About This Item
Recommended Products
form
viscous liquid
mol wt
average Mn ~2,000
refractive index
n20/D 1.451
density
0.996 g/mL at 25 °C
SMILES string
CC(N)CO.CC(O)CO
InChI
1S/C4H9NO2/c6-3-1-5-2-4(3)7/h3-7H,1-2H2
InChI key
JCZPOYAMKJFOLA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 3 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
365.0 °F - closed cup
Flash Point(C)
185 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service