Skip to Content
Merck
All Photos(1)

Documents

406449

Sigma-Aldrich

Tricosafluorododecanoic acid

95%

Synonym(s):

Perfluorododecanoic acid, Perfluorolauric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)10CO2H
CAS Number:
Molecular Weight:
614.10
Beilstein:
1811257
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

bp

245 °C/740 mmHg (lit.)

mp

105-108 °C (lit.)

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C12HF23O2/c13-2(14,1(36)37)3(15,16)4(17,18)5(19,20)6(21,22)7(23,24)8(25,26)9(27,28)10(29,30)11(31,32)12(33,34)35/h(H,36,37)

InChI key

CXGONMQFMIYUJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B O Fagbayigbo et al.
Environmental monitoring and assessment, 190(6), 346-346 (2018-05-17)
In this study, an analytical method for the routine determination of nine perfluorinated compounds (PFCs), using ultra performance liquid chromatography coupled to a quadrupole time-of-flight mass spectrometer (UPLC-QTOF-MS), was developed, validated, and used for their assay in surface water and
Lei Wang et al.
Journal of chromatography. A, 1515, 45-53 (2017-08-15)
Ionic liquids have been used to efficiently extract a wide range of polar and nonpolar organic contaminants from water. In this study, imidazole ionic liquids immobilized on silica gel were synthesized through a chemical bonding method, and the immobilized dodecylimidazolium
Wei Zhao et al.
Journal of environmental sciences (China), 56, 272-280 (2017-06-03)
Perfluoroalkyl substances (PFASs) are a class of fluorine substituted carboxylic acid, sulfonic acid and alcohol, structurally similar to their corresponding parent compounds. Previous study demonstrated the potential endocrine disruption and reproductive toxicity of perfluorooctane sulfonic acid and perfluorooctanoic acid, two
John L Newsted et al.
Environmental toxicology and chemistry, 36(11), 3138-3147 (2017-06-20)
In 2011, poly- and perfluoroalkyl substances (PFASs) were analyzed in surface water and fish fillet samples taken from Pool 2 of the Upper Mississippi River, a 33-mile stretch inclusive of the Minneapolis/St. Paul, Minnesota (USA) metropolitan area. Approximately 100 each
Changlong Wei et al.
Ecotoxicology and environmental safety, 152, 141-150 (2018-02-07)
Little research has been carried out for the per- and polyfluoroalkyl substances (PFASs) in groundwater from non-industrial areas, even though it has been proved that PFASs can transport for long distance. In this study, the concentration profiles and geographical distribution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service