Skip to Content
Merck
All Photos(3)

Key Documents

400173

Sigma-Aldrich

Lithium tert-butoxide

97%

Synonym(s):

LiOtBu

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COLi
CAS Number:
Molecular Weight:
80.05
Beilstein:
3620018
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder and chunks

bp

110  °C/at 0.1333 hPa

density

0,897 g/cm3

SMILES string

[Li+].CC(C)(C)[O-]

InChI

1S/C4H9O.Li/c1-4(2,3)5;/h1-3H3;/q-1;+1

InChI key

LZWQNOHZMQIFBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Lithium tert-butoxide (LiOtBu) is a weakly basic and nucleophilic alkali metal oxide commonly used as an initiator for anionic polymerization.
Other synthetic applications:
  • In combination with potassium diisopropylamide, LiOtBu can be used to deprotonate 1-(phenylseleno) alkenes and bis (phenylseleno) acetals.
  • LiOtBu can mediate the α-alkylation reaction of ketones with primary alcohols in the absence of any transition metal catalyst.
  • LiOtBu is an effective base for the synthesis of 3,4,5-trisubstituted 3H-oxazol-2-ones and 3,4-disubstituted (Z)-oxazolidin-2-ones from substituted propargyl alcohols and aryl/alkyl isocyanates using DMF as a solvent.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Self-heat. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lithium tert-Butoxide.
Caine D.
e-EROS Encyclopedia of Reagents for Organic Synthesis. null
LiOtBu Promoted 5?Exo?dig Cyclization of Propargyl Alcohols and Isocyanates for the Synthesis of Multisubstituted 3H?Oxazol?2?ones and Oxazolidin?2?ones.
Savarimuthu S A, et al.
ChemistrySelect, 1(9), 2035-2039 (2016)
The role of association/complexation equilibria in the anionic polymerization of (meth) acrylates.
Kunkel D, et al.
Macromolecular Symposia, 60(1), 315-326 (1992)
Organoselenium chemistry. 4. Deprotonations with potassium diisopropylamide-lithium tert-butoxide. Alkylation of 1-(phenylseleno) alkenes and bis (phenylseleno) acetals.
Raucher S and Koolpe G A
The Journal of Organic Chemistry, 43(19), 3794-3796 (1978)
Lithium tert-butoxide mediated α-alkylation of ketones with primary alcohols under transition-metal-free conditions.
Liang Y F, et al.
Royal Society of Chemistry Advances, 3(21), 7739-7742 (2013)

Articles

Few Monolayer Atomic Layer Deposition (ALD) on Surfaces and Interfaces for Energy Applications

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service