Skip to Content
Merck
All Photos(1)

Key Documents

396354

Sigma-Aldrich

Tetramethylgermanium

98%

Synonym(s):

Tetramethylgermane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4Ge
CAS Number:
Molecular Weight:
132.78
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

6.74 psi ( 20 °C)

Assay

98%

form

liquid

reaction suitability

core: germanium

refractive index

n20/D 1.389 (lit.)

bp

43-44 °C/740 mmHg (lit.)

mp

−88 °C (dec.) (lit.)

density

0.978 g/mL at 25 °C (lit.)

SMILES string

C[Ge](C)(C)C

InChI

1S/C4H12Ge/c1-5(2,3)4/h1-4H3

InChI key

ZRLCXMPFXYVHGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

CVD precursor to germanium oxide thin films.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-34.6 °F - closed cup

Flash Point(C)

-37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Thin Solid Films, 189, 293-293 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service