Skip to Content
Merck
All Photos(1)

Key Documents

388890

Sigma-Aldrich

Brij® S10

average Mn ~711

Synonym(s):

Polyethylene glycol octadecyl ether, Polyoxyethylene (10) stearyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H37(OCH2CH2)nOH, n~10
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mn ~711

mp

37-39 °C (lit.)

density

0.964 g/mL at 25 °C (lit.)

HLB

12

SMILES string

O(CC[O])CCCCCCCCCCCCCCCCCC

InChI

1S/C20H41O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-22-20-18-21/h2-20H2,1H3

InChI key

PSEGVHKUPXNGGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Brij is a registered trademark of Croda International PLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lingyan Song et al.
Journal of colloid and interface science, 328(2), 374-384 (2008-10-17)
Patternable nanoporous silica thin films with pore sizes on multiple length scales are fabricated using preformed block copolymer/homopolymer blend films as templates. Previous work by Tirumala et al. [V.R. Tirumala, R.A. Pai, S. Agarwal, J.J. Testa, G. Bhatnagar, A.H. Romang
Christian Klammt et al.
Journal of structural biology, 158(3), 482-493 (2007-03-14)
G-protein coupled receptors (GPCRs) are key elements in signal transduction pathways of eukaryotic cells and they play central roles in many human diseases. So far, most structural and functional approaches have been limited by the immense difficulties in the production
S Rahima Benhabbour et al.
Journal of controlled release : official journal of the Controlled Release Society, 158(1), 63-71 (2011-11-01)
Lipid-based oil-filled nanoparticles (NPs) with a high concentration of surface-chelated nickel (Ni-NPs) were successfully prepared using a Brij 78-NTA-Ni conjugate synthesized with Brij 78 (Polyoxyethylene (20) stearyl ether) and nitrilotriacetic acid (NTA). The facile incorporation of the Brij 78-NTA-Ni conjugate
Jae-Haeng Song et al.
Drug development and industrial pharmacy, 35(8), 897-903 (2009-02-26)
The oral administration of loratadine, an antihistamine, can have a variety of adverse side effects, such as headache, fatigue, and nausea, because of the transient high blood concentration. To avoid these effects, loratadine can be administered using a transdermal drug
J Q Zhang et al.
Drug delivery, 14(6), 381-387 (2007-08-19)
The study describes the development of stealth solid lipid nanoparticles (SLNs) as colloidal carriers for silibinin, a drug with very low solubility. Stealth SLNs were constituted mainly of bioacceptable and biodegradable lipids, such as stearic acid and surfactant Brij 78

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service