Skip to Content
Merck
All Photos(1)

Key Documents

383805

Sigma-Aldrich

1,4,8,11,15,18,22,25-Octabutoxy-29H,31H-phthalocyanine

Dye content 95 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H82N8O8
CAS Number:
Molecular Weight:
1091.38
Beilstein:
4289491
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

composition

Dye content, 95%

Quality Level

mp

221 °C (dec.) (lit.)

λmax

762 nm

SMILES string

CCCCOc1ccc(OCCCC)c2c3nc(nc4[nH]c(nc5nc(nc6[nH]c(n3)c7c(OCCCC)ccc(OCCCC)c67)c8c(OCCCC)ccc(OCCCC)c58)c9c(OCCCC)ccc(OCCCC)c49)c12

InChI

1S/C64H82N8O8/c1-9-17-33-73-41-25-26-42(74-34-18-10-2)50-49(41)57-65-58(50)70-60-53-45(77-37-21-13-5)29-30-46(78-38-22-14-6)54(53)62(67-60)72-64-56-48(80-40-24-16-8)32-31-47(79-39-23-15-7)55(56)63(68-64)71-61-52-44(76-36-20-12-4)28-27-43(75-35-19-11-3)51(52)59(66-61)69-57/h25-32H,9-24,33-40H2,1-8H3,(H2,65,66,67,68,69,70,71,72)

InChI key

KAPOSTKECKHKAP-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W E Ford et al.
Photochemistry and photobiology, 50(3), 277-282 (1989-09-01)
Triplet-state properties of 1,4,8,11,15,18,22,25-octa-n-butoxyphthalocyanine and its zinc derivative were determined for the first time. The T1 state of the metal-free phthalocyanine was characterized by a short lifetime (tau T = 17 microseconds) and low quantum yield (phi T = 0.095)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service