Skip to Content
Merck
All Photos(2)

Key Documents

379867

Sigma-Aldrich

Potassium dicyanoaurate(I)

99.95% trace metals basis

Synonym(s):

Gold (I) potassium cyanide, Potassium gold cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KAu(CN)2
CAS Number:
Molecular Weight:
288.10
Beilstein:
6235525
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.95% trace metals basis
form:
crystalline

Quality Level

Assay

99.95% trace metals basis

form

crystalline

impurities

≤550.0 ppm Trace Metal Analysis

density

3.45 g/mL at 25 °C (lit.)

SMILES string

[K+].N#C[Au-]C#N

InChI

1S/2CN.Au.K/c2*1-2;;/q;;-1;+1

InChI key

OQHPFUBKFKRHKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium dicyanoaurate(I) is an inorganic salt, soluble in water and sparingly soluble in alcohol. It is widely used as an electrolyte in the electrodeposition of gold on various substrates such as thin films and electrodes.

Application

Potassium dicyanoaurate(I) can be used:
  • To prepare gold nanowire array electrodes with fast charge transfer ability.
  • As a supporting electrolyte for the electrodeposition of gold in organic media.
  • To prepare polypyrrole-ClO4 gas sensors with fast response time and high sensitivity to volatile organic compounds.
  • In the electrochemical preparation of gold nanoparticle composites.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas sensing ability of a nanostructured conducting polypyrrole film prepared by catalytic electropolymerization on Cu/Au interdigital electrodes
Naader Alizadeh, et al.
Electroanalysis, 25, 2181-2192 (2013)
Gold electrodeposition in organic media
Lorena M.A. Monzon, et al.},
Journal of Electroanalytical Chemistry, 657, 54-60 (2011)
Effects of Nanowire Length on Charge Transport in Vertically Aligned Gold Nanowire Array Electrodes
Hideyuki Nakanishi, et al.
Langmuir, 34, 15674-15680 (2018)
Influence of substrate, additives, and pulse parameters on electrodeposition of gold nanoparticles from potassium Dicyanoaurate
Parisa Vahdatkhah and Sayed Khatiboleslam Sadrnezhaad
Metallurgical and Materials Transactions A: Physical Metallurgy and Materials Science, 46, 2584-2592 (2015)
R Silva et al.
Contact dermatitis, 37(2), 78-81 (1997-08-01)
During 1995, we added gold sodium thiosulfate and potassium dicyanoaurate to the patch test standard series. Our purpose was to compare the reactivity of the gold salts, to determine the frequency of contact allergy to gold in our country, to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service