Skip to Content
Merck
All Photos(1)

Key Documents

373761

Sigma-Aldrich

2,2,2-Trifluoroethyl methacrylate

contains 50-200 ppm MEHQ as inhibitor, 99%

Synonym(s):

2,2,2-Trifluoroethyl 2-methylprop-2-enoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2CH2CF3
CAS Number:
Molecular Weight:
168.11
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

liquid

contains

50-200 ppm MEHQ as inhibitor

refractive index

n20/D 1.361 (lit.)

bp

59 °C/100 mmHg (lit.)

density

1.181 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCC(F)(F)F

InChI

1S/C6H7F3O2/c1-4(2)5(10)11-3-6(7,8)9/h1,3H2,2H3

InChI key

QTKPMCIBUROOGY-UHFFFAOYSA-N

General description

2,2,2-Trifluoroethyl methacrylate (TFEMA) is a semifluorinated monomer that can be synthesized from methacryloyl chloride and 2,2,2-trifluoroethanol in presence of triethylamine. Its properties include chemical inertness, good wear resistance and low dielectric constant.

Application

TFEMA can be used in the preparation of poly(TFEMA), which can be used in the development of acrylic protective coatings.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Photochemical stability of partially fluorinated acrylic protective coatings I. Poly (2, 2, 2-trifluoroethyl methacrylate) and poly (1H, 1H, 2H, 2H-perfluorodecyl methacrylate-co-2-ethylhexyl methacrylate) s
Chiantore O, et al.
Polymer Degradation and Stability, 67(3), 461-467 (2000)
Yoshinori Kadoma
Dental materials journal, 29(5), 602-608 (2010-09-10)
The kinetic polymerization behavior of 2,2,2-trifluoroethyl methacrylate (TFEMA), 1,1,1,3,3,3-hexafluoroisopropyl methacrylate (HFIPMA), 2,2,2-trifluoroethyl acrylate (TFEA) and 1,1,1,3,3,3-hexafluoroisopropyl acrylate (HFIPA) was determined by isothermal differential scanning calorimetry (DSC) and high-performance liquid chromatography (HPLC) in order to improve the properties of fluorinated powder-liquid
Grafting modification of ramie fibers with poly (2, 2, 2-trifluoroethyl methacrylate) via reversible addition-fragmentation chain transfer (RAFT) polymerization in supercritical carbon dioxide
Liu X, et al.
Reactive and Functional Polymers, 70(12), 972-979 (2010)
Yoshinori Kadoma et al.
Dental materials journal, 28(5), 642-648 (2009-10-14)
In order to clarify the effect of fluorination of an adhesive resin on the durability of the resin bond to precious metal alloys, 2,2,2-trifluoroethyl methacrylate (TFEMA)-poly(2,2,2-trifluoroethyl methacrylate) (PTFEMA)/TBBO adhesive resin was prepared. The tensile bond strength of this resin to
Arda Yurtsever et al.
Electrophoresis, 30(4), 589-598 (2009-01-22)
A new, fluorinated monolithic stationary phase for CEC was first synthesized by a single-stage, thermally initiated copolymerization of a fluorinated monomer, 2,2,2-trifluoroethyl methacrylate (TFEM) and ethylene dimethacrylate (EDMA) in the presence of a porogen mixture. In this preparation, 2-acrylamido-2-methyl-1-propanesulfonic acid

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service