Skip to Content
Merck
All Photos(2)

Key Documents

368113

Sigma-Aldrich

3,5-Dihydroxybenzaldehyde

98%

Synonym(s):

α-Resorcylaldehyde (6CI)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CHO
CAS Number:
Molecular Weight:
138.12
Beilstein:
1930147
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

153-158 °C (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)c1cc(O)cc(O)c1

InChI

1S/C7H6O3/c8-4-5-1-6(9)3-7(10)2-5/h1-4,9-10H

InChI key

HAQLHRYUDBKTJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Condensation of isopropyl ethers of p-hydroxyphenyl acetic acid and 3,5-dihydroxybenzaldehyde has been reported.

Application

3,5-Dihydroxybenzaldehyde is suitable for use in the synthesis of 3,5-dihydroxyphenylglycine (3,5-DHPG). It may be used in the synthesis of:
  • fulgide7
  • 3,5-bis(undeca-4,6-diynyloxy)benzaldehyde
  • 3,5-didodecyloxybenzaldehyde
  • 1′-methyl-1′,5′-dihydro-2′-(3,5-bis(undeca-4,6-diynyloxy)phenyl)-1H-pyrrolo[3′,4′:1,9](C60-Ih)[5,6]fullerene (F2D)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denis E Markov et al.
The journal of physical chemistry. A, 109(24), 5266-5274 (2006-07-15)
Exciton diffusion and photoluminescence quenching in conjugated polymer/fullerene heterostructures are studied by time-resolved photoluminescence. It is observed that heterostructures consisting of a spin-coated poly(p-phenylene vinylene) (PPV)-based derivative and evaporated C60 are ill-defined because of diffusion of C60 into the polymer
Ivan K Ilic et al.
ChemSusChem, 13(7), 1856-1863 (2020-02-07)
Although several recent publications describe cathodes for electrochemical energy storage materials made from regrown biomass in aqueous electrolytes, their transfer to lithium-organic batteries is challenging. To gain a deeper understanding, we investigate the influences on charge storage in model systems
Enzymatic resolution and pharmacological activity of the enantiomers of 3, 5-dihydroxyphenylglycine, a metabotropic glutamate receptor agonist.
Richard BS, et al.
Bioorganic & Medicinal Chemistry Letters, 5(3), 223-228 (1995)
A re-investigation of resveratrol synthesis by Perkins reaction. Application to the synthesis of aryl cinnamic acids.
Solladie G, et al.
Tetrahedron, 59(18), 3315-3321 (2003)
A Porel et al.
Indian journal of pharmaceutical sciences, 73(1), 46-56 (2011-12-02)
The aim of the present study was the development and subsequent validation of a simple, precise and stability-indicating reversed phase HPLC method for the simultaneous determination of guaifenesin, terbutaline sulphate and bromhexine hydrochloride in the presence of their potential impurities

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service