Skip to Content
Merck
All Photos(1)

Key Documents

366897

Sigma-Aldrich

Palmitic acid-d31

98 atom % D, 99% (CP)

Synonym(s):

Hexadecanoic-d31 acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)14CO2H
CAS Number:
Molecular Weight:
287.62
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Assay

99% (CP)

form

solid

mp

61-64 °C

mass shift

M+31

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(O)=O

InChI

1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2

InChI key

IPCSVZSSVZVIGE-SAQPIRCFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Palmitic acid-d31 is an isotopic analogue of palmitic acid.

Application

Palmitic acid-d31 can be used in the synthesis of saturated and unsaturated fatty acids. It is used in quantitative determination of fatty acids from plastic labware.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service