Skip to Content
Merck
All Photos(2)

Key Documents

365734

Sigma-Aldrich

2-Amino-4-(trifluoromethyl)benzenethiol hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(CF3)SH · HCl
CAS Number:
Molecular Weight:
229.65
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

196-198 °C (lit.)

functional group

fluoro

SMILES string

Cl.Nc1cc(ccc1S)C(F)(F)F

InChI

1S/C7H6F3NS.ClH/c8-7(9,10)4-1-2-6(12)5(11)3-4;/h1-3,12H,11H2;1H

InChI key

FIAGYDIJZOWVAB-UHFFFAOYSA-N

Application

2-Amino-4-(trifluoromethyl)benzenethiol hydrochloride may be used in the synthesis of 6-trifluoromethyl-4H-1,4-benzothiazines, via condensation and oxidative cyclization with β-di-carbonyl compounds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 6-trifluoromethyl-4h-1, 4-benzothiazines as possible anticancer agents.
Gupta RR and Kumar R.
Journal of Fluorine Chemistry, 31(1), 19-24 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service