Skip to Content
Merck
All Photos(1)

Key Documents

349410

Sigma-Aldrich

Zinc

foil, thickness 1.0 mm, 99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
EC Number:
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 487 °C)

Quality Level

Assay

99.99% trace metals basis

form

foil

resistivity

5.8 μΩ-cm, 20°C

thickness

1.0 mm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Quantity

25×25 mm (approximately 4.5 g)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Zinc foil, 6mm disks, thickness 1.0mm, as rolled, 99.95+%

GF32323339

Zinc

K(8)Tl(10)Zn: A Zintl Phase Containing the Zinc-Centered Thallium Polyanion Tl(10)Zn(8)(-).
Zhen-Chao Dong et al.
Inorganic chemistry, 36(16), 3559-3561 (1997-07-30)
M E Kuyumcu et al.
European review for medical and pharmacological sciences, 17(11), 1467-1471 (2013-06-19)
Zinc is one of the most important elements for human body. Zinc deficiency can occur in any age, if it is seen in elderly its clinical results can be more harmful due to already diminished functions. Some studies showed zinc
Sara Eyal et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 50(5), 798-806 (2009-05-01)
Studies in rodents indicate that the disruption of P-glycoprotein (P-gp) function increases drug distribution into the developing fetus and organs such as the brain. To simultaneously and serially evaluate the effect of P-gp activity and inhibition on the tissue distribution
W Reainthippayasakul et al.
Journal of nanoscience and nanotechnology, 13(5), 3617-3621 (2013-07-19)
Conjugated meso-alkynyl 5,15-dimesitylporphyrin metal complexes have been synthesized by Sonogashira coupling reaction in good yields. Alkynyl groups were chosen as a link at the meso positions in order to extend the pi-conjugated length of porphyrin rings. These synthesized porphyrin derivatives
Lena Martin et al.
Archives of animal nutrition, 67(3), 248-261 (2013-06-08)
This study aimed at investigating the effect of dietary zinc on performance, jejunal brush border membrane enzyme activities and mRNA levels of enzymes and two zinc transporters in piglets. A total of 126 piglets were weaned at 26 ±1 days

Articles

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service