Skip to Content
Merck
All Photos(3)

Key Documents

345857

Sigma-Aldrich

Tetrabutylammonium chloride hydrate

98%

Synonym(s):

TBAC hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(Cl) · xH2O
CAS Number:
Molecular Weight:
277.92
Beilstein:
3765327
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

41-44 °C (lit.)

SMILES string

[Cl-].[N+](CCCC)(CCCC)(CCCC)CCCC.O

InChI

1S/C16H36N.ClH.H2O/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;;/h5-16H2,1-4H3;1H;1H2/q+1;;/p-1

InChI key

FODWRUPJCKASBN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Tetrabutylammonium chloride (TBAC) hydrate has been used as a phase transfer catalyst in the synthesis of fullerenols and in Heck-type palladium-catalyzed vinylation of organic halides in water. TBAC hydrate has also been reported to be a promising candidate for cool energy storage material.
Tetrabutylammonium chloride hydrate can be used:
  • As an additive in palladium-catalyzed cross-coupling reaction of heteroaromatic carboxylic acids with aryl bromides.
  • To enhance the reactivity of palladium-catalyzed arylation of methyl acrylates.
  • In derivatization of bis(spirodienone) calixarene having exocyclic double bonds to its chloro derivative.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phase equilibrium of ionic semiclathrate hydrates formed with tetrabutylammonium bromide and tetrabutylammonium chloride.
Sato K, et al.
Fluid Phase Equilibria, 337, 115-118 (2013)
Synthesis of p-tert-butylcalix-4-arene derivatives with trans-alkyl substituents on opposite methylene bridges
Simaan S and Biali SE
The Journal of Organic Chemistry, 68(9), 3634-3639 (2003)
Unexpected intermolecular Pd-catalyzed cross-coupling reaction employing heteroaromatic carboxylic acids as coupling partners
Forgione P, et al.
Journal of the American Chemical Society, 128(35), 11350-11351 (2006)
On the efficiency of tetraalkylammonium salts in Heck type reactions
Jeffery T
Tetrahedron, 52(30), 10113-10130 (1996)
Electrical and optical properties of fullerenol Langmuir? Blodgett films deposited on polyaniline substrates.
Rincon M E, et al.
The Journal of Physical Chemistry B, 107(17), 4111-4117 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service