Skip to Content
Merck
All Photos(2)

Key Documents

345415

Sigma-Aldrich

N-Methoxy-N-methylbenzamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CON(OCH3)CH3
CAS Number:
Molecular Weight:
165.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.533 (lit.)

bp

70 °C/0.1 mmHg (lit.)

density

1.085 g/mL at 25 °C (lit.)

functional group

amine
phenyl

SMILES string

CON(C)C(=O)c1ccccc1

InChI

1S/C9H11NO2/c1-10(12-2)9(11)8-6-4-3-5-7-8/h3-7H,1-2H3

InChI key

UKERDACREYXSIV-UHFFFAOYSA-N

General description

N-Methoxy-N-methylbenzamide is an N,N-disubstituted benzamide. also referred as Weinreb amide., Hydrogen bonding interactions between thioacetamide and N-methoxy-N-methylbenzamide has been investigated using near-infrared absorption spectroscopy. Preparation of N-methoxy-N-methylbenzamide has been reported.

Application

N-Methoxy-N-methylbenzamide may be used in the preparation of β-trifluoromethyl enaminones.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Near-infrared spectroscopic studies of the hydrogen bonding between thioacetamide and< i> N</i>,< i> N</i>-disubstituted benzamide derivatives in CCl< sub> 4</sub>.
Choi YS, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 52(13), 1779-1783 (1996)
An efficient conversion of carboxylic acids into Weinreb amides.
Katritzky AR, et al.
ARKIVOC (Gainesville, FL, United States), 11, 39-44 (2002)
A novel approach to ?-trifluoromethyl enaminones.
Jeong IH, et al.
Tetrahedron Letters, 43(40), 7171-7174 (2002)
Kazuhito Hioki et al.
Chemical & pharmaceutical bulletin, 52(4), 470-472 (2004-04-02)
Weinreb amides were successfully prepared from the corresponding carboxylic acids using 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride (DMT-MM) in the solvents, methanol, isopropyl alcohol, and acetonitrile, which can solubilize DMT-MM. A variety of carboxylic acids were converted to the corresponding Weinreb amides in excellent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service