Skip to Content
Merck
All Photos(2)

Documents

344214

Sigma-Aldrich

2-Bromo-3′-nitroacetophenone

97%

Synonym(s):

3′-Nitrophenacyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4COCH2Br
CAS Number:
Molecular Weight:
244.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

90-96 °C (lit.)

SMILES string

[O-][N+](=O)c1cccc(c1)C(=O)CBr

InChI

1S/C8H6BrNO3/c9-5-8(11)6-2-1-3-7(4-6)10(12)13/h1-4H,5H2

InChI key

GZHPNIQBPGUSSX-UHFFFAOYSA-N

General description

Debromination of 2-bromo-3′-nitroacetophenone in various solvents has been reported.

Application

2-Bromo-3′-nitroacetophenone may be used in the preparation of 2-bromo-3′-nitroacetophenone. It may be used in the preparation of 2-hydroxy-ethyl-1-[(3-nitro-phenyl)-2-oxoethyl]-piperidinium bromide.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Debromination of a-Bromoketones and vic-Dibromides Using a NaI/Na2SO3 System.
Lee SH, et al.
Bull. Korean Chem. Soc., 25(11), 1723-1723 (2004)
Sarwat Jahan et al.
Pakistan journal of pharmaceutical sciences, 26(3), 517-523 (2013-04-30)
Synthesis of novel phenacyl derivatives of alkyl piperidine as cytotoxic agents via simple and single step reaction procedure is going to be reported here. Twelve new compounds were successfully synthesized in moderate yield and in solid form. Their synthesis was
3-Aryl-1, 2-dihydroquinoxalines.
Figueras J.
The Journal of Organic Chemistry, 31(3), 803-806 (1966)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service